Skip to Content
Merck
All Photos(1)

Key Documents

N9037

Sigma-Aldrich

Necrostatin-1

≥98% (HPLC), powder, necroptosis inhibitor

Synonym(s):

5-(1H-Indol-3-ylmethyl)-3-methyl-2-thioxo-4-Imidazolidinone, 5-(Indol-3-ylmethyl)-3-methyl-2-thio-Hydantoin, MTH-DL-Tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3OS
CAS Number:
Molecular Weight:
259.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Necrostatin-1, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

CN1C(=S)NC(Cc2c[nH]c3ccccc23)C1=O

InChI

1S/C13H13N3OS/c1-16-12(17)11(15-13(16)18)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,14H,6H2,1H3,(H,15,18)

InChI key

TXUWMXQFNYDOEZ-UHFFFAOYSA-N

Application

Necrostatin-1has been used to investigate its effect on the regulation of interleukin (IL)-33 expression during liver injury in mice in vivo and in hepatic stellate cells in vitro. It has also been used as an inhibitor of receptor-interacting protein kinase 1 (RIPK1) in experimental studies.

Biochem/physiol Actions

Necrostatin-1 is an inhibitor of necroptosis (non-apoptotic cell death pathway).
Necrostatin-1(Nec-1) might exhibit therapeutic benefit against patients with traumatic brain injury (TBI). In addition, it might also be used to treat other acute central nervous system (CNS) disorders that feature necroptosis as a mode of cell death. It is extensively used to study the effect of receptor-interacting protein kinase 1 (RIPK1) on cell death and inflammation in experimental disease models. Nec‐1 protects hippocampal (HT‐22) cells from glutamate‐induced oxytosis via increasing cellular glutathione (GSH) levels, decreasing reactive oxygen species production, inhibiting the nuclear translocation of apoptosis‐inducing factor and B-cell lymphoma 2 (Bcl‐2) /adenovirus E1B 19kDa‐interacting protein 3‐related pathways.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anahita Kyani et al.
ChemMedChem, 13(2), 164-177 (2017-12-14)
Protein disulfide isomerase (PDI) is overexpressed in glioblastoma, the most aggressive form of brain cancer, and folds nascent proteins responsible for the progression and spread of the disease. Herein we describe a novel nanomolar PDI inhibitor, pyrimidotriazinedione 35G8, that is
Necrostatin-1 analogues: critical issues on the specificity, activity and in vivo use in experimental disease models
Takahashi N, et al.
Cell Death & Disease, 3(11), e437-e437 (2012)
Necrostatin-1 reduces histopathology and improves functional outcome after controlled cortical impact in mice
You Z, et al.
Journal of Cerebral Blood Flow and Metabolism, 28(9), 1564-1573 (2008)
Sorafenib-induced defective autophagy promotes cell death by necroptosis
Kharaziha P, et al.
Testing, 6(35), 37066-37066 (2015)
T Suzuki et al.
Canadian journal of biochemistry, 55(5), 521-527 (1977-05-01)
Proton magnetic resonance spectra of methylthiohydantoin (3-methyl-2-thiohydantoin), thiohydantoin (2-thiohydantoin), and hydantoin derivatives of amino acids were studied in dimethyl sulfoxide-d6. Their parent amino acids could be identified by the spectra. An application to the N- and C-terminal amino acid analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service