Skip to Content
Merck
All Photos(1)

Documents

SML1603

Sigma-Aldrich

BAY-598

≥98% (HPLC)

Synonym(s):

(S)-N-[1-{N′-Cyano-N-[3-(difluoromethoxy)phenyl]carbamimidoyl}-3-(3,4-dichlorophenyl)-4,5-dihydro-1H-pyrazol-4-yl]-N-ethyl-2-hydroxyacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20Cl2F2N6O3
Molecular Weight:
525.34
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -85 to -95°, c = 1 in methanol

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C=C(C2=NN(/C(NC3=CC=CC(OC(F)F)=C3)=N\C#N)C[C@@H]2N(CC)C(CO)=O)C=C1

InChI

1S/C23H21Cl2F2N5O3/c1-2-16(20(34)11-33)17-10-32(31-21(17)13-6-7-18(24)19(25)8-13)23(29-12-28)30-14-4-3-5-15(9-14)35-22(26)27/h3-9,16-17,22,33H,2,10-11H2,1H3,(H,29,30)/t16?,17-/m1/s1

InChI key

LOHNYXGTWKNYDF-ZYMOGRSISA-N

Biochem/physiol Actions

BAY-598 is a potent, peptide-competitive chemical probe for SET and MYND domain-containing protein 2 (SMYD2), a lysine methyl transferase inhibitor that dimethylates histone H3K36 and methylates histone H3K4. SMYD2 also methylates Lys-370 of p53, leading to decreased DNA-binding activity. SMYD2 is over-expressed in several cancers with poor prognosis. BAY-598 inhibits in vitro methylation of p53K370 with an IC50 value of 27 nM and in cells with an IC50 value < 1 μM. BAY-598 is more than 100-fold selective over other histone methyltransferases and non-epigenetic targets. BAY-598 can be used with in vivo experiments. For full characterization details, please visit the BAY-598 probe summary on the Structural Genomics Consortium (SGC) website.

BAY-369 is the negative control for the active probe, BAY-598. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

BAY-598 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

BAY-598 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the BAY-598 probe summary on the Chemical Probes Portal website.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenchuan Liu et al.
Clinical and translational medicine, 11(9), e545-e545 (2021-09-30)
Esophageal squamous cell carcinoma (ESCC) is a highly aggressive and treatment-resistant tumor. The biological implications and molecular mechanism of cancer stem-like cells (CSCs) in ESCC, which contribute to therapeutic resistance such as radioresistance, remain elusive. Quantitative real-time polymerase chain reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service