Skip to Content
Merck
All Photos(1)

Key Documents

SML1546

Sigma-Aldrich

SCR7 pyrazine

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-6,7-diphenyl-2-thioxo-4(1H)-pteridinone, 6,7-Diphenyl-2-thio-lumazine (8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N4OS
CAS Number:
Molecular Weight:
332.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(C1=C(N2)N=C(C3=CC=CC=C3)C(C4=CC=CC=C4)=N1)NC2=S

InChI

1S/C18H12N4OS/c23-17-15-16(21-18(24)22-17)20-14(12-9-5-2-6-10-12)13(19-15)11-7-3-1-4-8-11/h1-10H,(H2,20,21,22,23,24)

InChI key

GSRTWXVBHGOUBU-UHFFFAOYSA-N

General description

SCR7 pyrazine is an inhibitor of DNA ligase IV.

Application

SCR7 pyrazine has been used as a non-homologous end joining (NHEJ) modulator to study its effect on CRISPR/Cas9-mediated editing.
SCR7 pyrazine has been shown to enhance CRISPR genome editing efficiency. To see other small molecule CRISPR enhancers, visit sigma.com/CRISPR-enhancers.

Biochem/physiol Actions

SCR7 pyrazine is reported to be an inhibitor of non-homologous end joining (NHEJ) and has been shown to enhance the efficiency of CRISPR-Cas9 genome editing.
SCR7 pyrazine is reported to be an inhibitor of non-homologous end joining (NHEJ) and has been shown to enhance the efficiency of CRISPR-Cas9 genome editing. The effect of SCR7 pyrazine on the efficiency and targeting precision of CRISPR applications has been shown to be cell type specific and context dependent. SCR7 pyrazine is a product of spontaneous cyclization of SCR7, first reported by Srivastava, M., et al.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

pRB-Depleted Pluripotent Stem Cell Retinal Organoids Recapitulate Cell State Transitions of Retinoblastoma Development and Suggest an Important Role for pRB in Retinal Cell Differentiation.
Rozanska, et al.
Stem Cells Translational Medicine, 11, 415-433 (2023)
Diane Yang et al.
Scientific reports, 6, 21264-21264 (2016-02-19)
Efficient gene editing is essential to fully utilize human pluripotent stem cells (hPSCs) in regenerative medicine. Custom endonuclease-based gene targeting involves two mechanisms of DNA repair: homology directed repair (HDR) and non-homologous end joining (NHEJ). HDR is the preferred mechanism
Disruption of diphthamide synthesis genes and resulting toxin resistance as a robust technology for quantifying and optimizing CRISPR/Cas9-mediated gene editing.
Killian T, et al.
Scientific Reports, 7(1), 15480-15480 (2017)
Justin Moser et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(35), E8219-E8227 (2018-08-17)
The Restriction Point was originally defined as the moment that cells commit to the cell cycle and was later suggested to coincide with hyperphosphorylation of the retinoblastoma protein (Rb). Current cell cycle models posit that cells exit mitosis into a
Yuanwu Ma et al.
RNA biology, 13(7), 605-612 (2016-05-11)
Precise modifications such as site mutation, codon replacement, insertion or precise targeted deletion are needed for studies of accurate gene function. The CRISPR/Cas9 system has been proved as a powerful tool to generate gene knockout and knockin animals. But the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service