Skip to Content
Merck
All Photos(2)

Documents

SML1229

Sigma-Aldrich

2′,3′-cGAMP sodium salt

≥98% (HPLC)

Synonym(s):

2′,5′-3′,5′-cGAMP, Cyclic [G(2′,5′)pA(3′,5′)p], cGAMP(2′-5′), c[G(2′,5′)pA(3′,5′)p] sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N10O13P2 · xNa+
CAS Number:
Molecular Weight:
674.41 (free acid basis)
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

description

contains 0.5μmol of powder (approx. 0.335mg)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

O=C1N=C(N)NC2=C1N=CN2[C@H](O3)[C@H](OP(OC[C@@H]4[C@@H](O5)[C@@H](O)[C@H](N6C=NC7=C6N=CN=C7N)O4)(O)=O)[C@H](O)[C@H]3COP5(O)=O

InChI

1S/C20H24N10O13P2/c21-14-8-15(24-3-23-14)29(4-25-8)18-11(32)12-7(41-18)2-39-45(36,37)43-13-10(31)6(1-38-44(34,35)42-12)40-19(13)30-5-26-9-16(30)27-20(22)28-17(9)33/h3-7,10-13,18-19,31-32H,1-2H2,(H,34,35)(H,36,37)(H2,21,23,24)(H3,22,27,28,33)/t6-,7-,10-,11-,12-,13-,18-,19-/m1/s1

InChI key

XRILCFTWUCUKJR-INFSMZHSSA-N

Application

2′,3′-cGAMP sodium salt has been used to identify small compounds capable of binding human stimulator of interferon genes (STING). It is also used to study type I interferon response to cytosolic DNA.

Biochem/physiol Actions

2′,3′-cGAMP (cyclic [G(2′,5′)pA(3′,5′)p]) is the natural agonist for the STING (STimulator of INterferon Genes) pathway, discovered to be a pathway for antiviral innate immunity. 2′3′-cGAMP is a second messenger produced in mammalian cells by the cytosolic DNA sensor cGAS (cGAMP synthase) to activate innate immune responses by binding to STING and initiating an interferon response. The affinity of 2′3′-cGAMP for human STING is much higher than that of the bacterial 3′3′-cyclic dinucleotides (CDNs), with a Kd of 4.59 nM compared to >1 μM for bacterial 3′3′-CDNs.

Preparation Note

2′,3′-cGAMP is soluble in water and aqueous buffers (>> 5 mM, limits have not been determined).

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua-Fang Bao et al.
The FEBS journal, 291(15), 3432-3453 (2024-04-28)
Inflammatory signals from immunological cells may cause damage to intestinal epithelial cells (IECs), resulting in intestinal inflammation and tissue impairment. Interferon-γ-inducible protein 16 (IFI16) was reported to be involved in the pathogenesis of Behçet's syndrome (BS). This study aimed to
Qingyuan Zhu et al.
Antiviral research, 187, 105015-105015 (2021-01-15)
The newly emerged severe acute respiratory syndrome-coronavirus-2 (SARS-CoV-2) coronavirus initiated a pneumonia outbreak (COVID-19) that rapidly spread worldwide and quickly became a public health emergency of international concern; However to date, except Remdesivir, there are no clinically approved specific or
Christos Dogrammatzis et al.
mBio, 15(4), e0037324-e0037324 (2024-03-12)
The STimulator of INterferon Genes (STING) constitutes a major DNA-sensing pathway that restricts HSV-1 infection in different models by activating type I interferon and pro-inflammatory responses. To counteract STING, HSV-1 has evolved numerous strategies including mechanisms to interfere with its
Qingyuan Zhu et al.
Antiviral research, 183, 104933-104933 (2020-09-20)
Stimulator of interferon genes (STING), as a signaling hub in innate immunity, plays a central role for the effective initiation of host defense mechanisms against microbial infections. Upon binding of its ligand cyclic dinucleotides (CDNs) produced by the cyclic GMP-AMP
Cuicui Ma et al.
Signal transduction and targeted therapy, 6(1), 353-353 (2021-10-02)
Pseudomonas aeruginosa infection continues to be a major threat to global public health, and new safe and efficacious vaccines are needed for prevention of infections caused by P. aeruginosa. X-ray irradiation has been used to prepare whole-cell inactivated vaccines against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service