Skip to Content
Merck
All Photos(1)

Key Documents

F3503

Sigma-Aldrich

5-Fluoro-2′-deoxyuridine 5′-monophosphate sodium salt

~85%

Synonym(s):

FdUMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN2O8P
CAS Number:
Molecular Weight:
326.17
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~85%

form

solid

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(O)=O)N2C=C(F)C(=O)NC2=O

InChI

1S/C9H12FN2O8P.Na.H/c10-4-2-12(9(15)11-8(4)14)7-1-5(13)6(20-7)3-19-21(16,17)18;;/h2,5-7,13H,1,3H2,(H,11,14,15)(H2,16,17,18);;

InChI key

DGEOOGBABXVZPJ-UHFFFAOYSA-N

General description

5- 5-fluoro-2′-deoxyuridylate (FdUMP) is an intracellularly generated metabolite of 5-Fluorouracil (5-FU), a widely used reagent in molecular biology to study DNA synthesis and metabolism.

Application

It has been used:

  • as an anti-mitotic agent for culturing mouse primary cortical neurons.
  • as a component of feeding media in the culturing of mouse primary cortical neurons.
  • in studies on the activity and function of thymidylate synthase (TS).

Biochem/physiol Actions

5-Fluoro-2′-deoxyuridine 5′-monophosphate sodium salt (FdUMP) is an DNA synthesis inhibitor. FdUMP along with RNA synthesis inhibitor 3′-C-ethinylcytidine induces cell death in cervical cancer cells at nanomolar concentrations.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yoshihiro Nabeya et al.
Cancer science, 102(8), 1509-1515 (2011-05-13)
Thymidylate synthase (TS) plays a major role in the response to 5-fluorouracil (5-FU) by binding directly to the 5-FU metabolite, 5-fluoro-dUMP (FdUMP). The change in the TS expression levels after 5-FU administration was examined in parallel to 5-FU responsiveness in
Comparison of partially and fully chemically-modified siRNA in conjugate-mediated delivery in vivo
Hassler MR, et al.
Nucleic Acids Research, 46(5), 2185-2196 (2018)
Hydrophobicity of Lipid-Conjugated siRNAs Predicts Productive Loading to Small Extracellular Vesicles
Biscans A, et al.
Molecular Therapy, 26(6), 1520-1528 (2018)
A high-throughput assay for mRNA silencing in primary cortical neurons in vitro with oligonucleotide therapeutics
Alterma n JF, et al.
Bio-protocol, 7(16) (2017)
Adam Jarmuła et al.
Bioorganic & medicinal chemistry letters, 18(8), 2701-2708 (2008-03-26)
Molecular dynamics simulations and free energy calculations are presented, exploring previously described experimentally studied interactions of a series of 2'-fluoro-substituted dUMP/FdUMP analogues with thymidylate synthase (TS). The results show the inhibitory behaviors of 2'-F-ara-UMP, 2',2''-diF-dUMP and 2',5-diF-ara-UMP to be dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service