Skip to Content
Merck
All Photos(1)

Documents

SML0902

Sigma-Aldrich

MONNA

≥98% (HPLC)

Synonym(s):

N-((4-Methoxy)-2-naphthyl)-5-nitroanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O5
Molecular Weight:
338.31
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MONNA (benzbromarone (Benzb) and N-((4-methoxy)-2-naphthyl)-5-nitroanthranilic acid) is a TMEM16A antagonists. It affects the liberation of sarcoplasmic reticulum (SR) calcium in human airway smooth muscle cells.
MONNA is a TMEM16A (ANO1) calcium-activated chloride channel blocker. MONNA blocks rhTMEM16A (ANO1) channel currents in HEK293 cells with and IC50 of 1.27 μM. The compound MONNA does not affect related human or mouse chloride channels bestrophin-1, CLC2 and CFTR at concentrations up to 10 μM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antagonists of the TMEM16A calcium-activated chloride channel modulate airway smooth muscle tone and intracellular calcium.
Danielsson J, et al.
Anesthesiology, 123(3), 569-581 (2015)
Quan-Fang Zhang et al.
Frontiers in plant science, 11, 145-145 (2020-03-13)
Ceramide sphingolipids are major components of membranes. C2 and C6 ceramides induce programmed cell death (PCD) in animals and plants, and we previously showed that C2 and C6 ceramides induce PCD in rice (Oryza sativa) protoplasts. However, the mechanistic link
Hayley Pearson et al.
Thorax, 76(1), 64-72 (2020-10-29)
Human respiratory syncytial virus (HRSV) is a common cause of respiratory tract infections (RTIs) globally and is one of the most fatal infectious diseases for infants in developing countries. Of those infected, 25%-40% aged ≤1 year develop severe lower RTIs leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service