Merck
  • Ceramide-Induced Cell Death Depends on Calcium and Caspase-Like Activity in Rice.
Ceramide-Induced Cell Death Depends on Calcium and Caspase-Like Activity in Rice.
Frontiers in plant science (2020-03-13)
Quan-Fang Zhang, Jian Li, Fang-Cheng Bi, Zhe Liu, Zhen-Yi Chang, Ling-Yan Wang, Li-Qun Huang, Nan Yao
ABSTRACT
Ceramide sphingolipids are major components of membranes. C2 and C6 ceramides induce programmed cell death (PCD) in animals and plants, and we previously showed that C2 and C6 ceramides induce PCD in rice (Oryza sativa) protoplasts. However, the mechanistic link between sphingolipids and PCD in rice remains unclear. Here, we observed that calcium levels increased rapidly after ceramide treatment. Moreover, the calcium channel inhibitor LaCl3 and the intracellular calcium chelator acetoxymethyl-1, 2-bis (2-aminophenoxy) ethic acid (BAPTA-AM) inhibited this calcium increase and prevented ceramide-induced PCD. Moreover, caspase-3-like protease activity increased significantly in C6 ceramide-treated protoplasts, and a caspase-specific inhibitor prevented C6 ceramide-induced cell death. We also detected the other typical PCD events including ATP loss. DIDS (4, 49-diisothiocyanatostilbene- 2, 29-disulfonic acid), an inhibitor of voltage-dependent anion channels (VDACs), decreased C6 ceramide-induced cell death. Together, this evidence suggests that mitochondria played an important role in C6 ceramide-induced PCD.
Sigma-Aldrich
N-acetylcysteine amide, ≥98% (HPLC)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
Glass beads, acid-washed, 425-600 μm (30-40 U.S. sieve)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Calcium Ionophore A23187 hemicalcium salt, powder
Sigma-Aldrich
N-Hexanoyl-D-sphingosine, ≥98% (TLC), synthetic
Sigma-Aldrich
MONNA, ≥98% (HPLC)