Skip to Content
Merck
All Photos(1)

Documents

M-168

Supelco

5-MeO-DMT solution

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N2O
CAS Number:
Molecular Weight:
218.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)CCC1=CNC2=CC=C(OC)C=C21

InChI

1S/C13H18N2O/c1-15(2)7-6-10-9-14-13-5-4-11(16-3)8-12(10)13/h4-5,8-9,14H,6-7H2,1-3H3

InChI key

ZSTKHSQDNIGFLM-UHFFFAOYSA-N

General description

5-MeO-DMT is a powerful, hallucinogenic tryptamine found in various plants used historically in the preparation of psychotropic snuffs. This certified solution standard is suitable for 5-MeO-DMT testing methods by GC/MS or LC/MS in clinical toxicology, urine drug testing, or forensic analysis applications.

Application


  • 5-MeO-DMT: Advancing Neuropharmacological Research.: As a potent research chemical, 5-MeO-DMT is pivotal in neuropharmacological studies, particularly those investigating the serotonergic system. Its profound effects on serotonin receptors offer valuable insights into mood regulation and cognitive processes, making it a critical tool for researchers studying psychiatric and neurological disorders (Dakic et al., 2017).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geovany Albino de Souza et al.
ACS omega, 5(15), 8804-8815 (2020-04-28)
In this work, a drug delivery system for perillyl alcohol based on the peptide self-assembly containing 3-(2-benzothiazolyl)-7-(diethylamino)coumarin (C6) as a fluorescent additive is obtained, and its photophysical characteristics as well as its release dynamics were studied by steady-state and time-resolved
Weipang Chang et al.
Nature communications, 12(1), 4857-4857 (2021-08-13)
Physical exercise stimulates adult neurogenesis, yet the underlying mechanisms remain poorly understood. A fundamental component of the innate neuroregenerative capacity of zebrafish is the proliferative and neurogenic ability of the neural stem/progenitor cells. Here, we show that in the intact
Byung Gon Jo et al.
Molecular medicine (Cambridge, Mass.), 26(1), 119-119 (2020-12-05)
Increasing number of studies provide evidence that the vagus nerve stimulation (VNS) dampens inflammation in peripheral visceral organs. However, the effects of afferent fibers of the vagus nerve (AFVN) on anti-inflammation have not been clearly defined. Here, we investigate whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service