Skip to Content
Merck
All Photos(1)

Key Documents

SML0680

Sigma-Aldrich

CYM-5541

≥98% (HPLC)

Synonym(s):

CYM5541, ML249, N,N-Dicyclohexyl(5-cyclopropylisoxazol-3-yl)-carboxamide, N,N-Dicyclohexyl-5-cyclopropyl-3-isoxazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28N2O2
CAS Number:
Molecular Weight:
316.44
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N(C4CCCCC4)(C3CCCCC3)C(=O)c1n[o]c(c1)C2CC2

InChI

1S/C19H28N2O2/c22-19(17-13-18(23-20-17)14-11-12-14)21(15-7-3-1-4-8-15)16-9-5-2-6-10-16/h13-16H,1-12H2

InChI key

NDKGACIWVAOUQH-UHFFFAOYSA-N

Biochem/physiol Actions

CYM-5541 is an allosteric agonist selective for the Sphingosine 1-phosphate (S1P) S1P3 receptor subtype. CYM-5541 occupies a different chemical space in the ligand binding pocket of S1P3 than S1P does, a lower hydrophobic region around transmembrane helix TM6. CYM-5541 shows great selectivity and stability with an EC50 of between 72 and 132 nM compared to S1P1 EC50 > 10 μM, S1P2 EC50 > 50 μM, S1P4 EC50 > 50 μM, and S1P5 EC50 > 25 μM and no significant activities for a panel of 55 GPCRs, ion channels, and transporters.

Features and Benefits

This compound is featured on the Lysophospholipid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service