Skip to Content
Merck
All Photos(1)

Key Documents

SML0460

Sigma-Aldrich

Methylhexanamine hydrochloride

≥98% (HPLC)

Synonym(s):

1,3-Dimethylamylamine hydrochloride, 1,3-Dimethylpentanamine hydrochloride, 1,3-Dimethylpentylamine hydrochloride, 2-Amino-4-methylhexane hydrochloride, 4-Methyl-2-hexylamine hydrochloride, DMAA, NSC 1106

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17N · HCl
CAS Number:
Molecular Weight:
151.68
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL (clear solution)

storage temp.

room temp

SMILES string

CCC(C)CC(C)N.[H]Cl

InChI

1S/C7H17N/c1-4-6(2)5-7(3)8/h6-7H,4-5,8H2,1-3H3

InChI key

YAHRDLICUYEDAU-UHFFFAOYSA-N

Biochem/physiol Actions

Methylhexanamine induces fat degradation and prevents energy loss during exercise. Thus, it serves as an active ingredient of many dietary supplements.
Methylhexanamine is a naturally substance isolated from Pelargonium graveolen. It is a component of geranium oil. Methylhexanamine is a sympathomimentic that increase levels of norepinephrine in the synaptic cleft.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxford Textbook of Children's Sport and Exercise Medicine (2017)
Hadeel A Alsufyani et al.
European journal of pharmacology, 843, 121-125 (2018-11-06)
We have investigated the mode of cardiovascular action of the stimulant methylhexaneamine (MHA) in terms of direct or indirect adrenergic actions in anaesthetised rats. Male and female rats were anaesthetised with pentobarbitone and pressor (changes in diastolic blood pressure) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service