Skip to Content
Merck
All Photos(1)

Documents

D2404

Sigma-Aldrich

Decylamine

95%

Synonym(s):

1-Aminodecane, NDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9NH2
CAS Number:
Molecular Weight:
157.30
Beilstein:
1735220
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

216-218 °C (lit.)

mp

12-14 °C (lit.)

density

0.787 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCN

InChI

1S/C10H23N/c1-2-3-4-5-6-7-8-9-10-11/h2-11H2,1H3

InChI key

MHZGKXUYDGKKIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Decylamine has been used as:
  • Amine in the synthesis of dynole-2-24 by reductive amination with 1-(3-(dimethylamino)propyl)-1H-indole-3-carbaldehyde.
  • A modifier in the synthesis of nanocomposites based on poly(butylene terephthalate) and C60 nanoparticles.
  • A template in the synthesis of ordered two and three dimensional mesoporous iron oxide.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Fullerene Derivates on Thermal and Crystallization Behavior of PBT/Decylamine-C60 and PBT/TCNEO-C60 Nanocomposites
Wozniak-Braszak A, et al.
Journal of Nanomaterials, (4) (2012)
Two-and Three-Dimensional Mesoporous Iron Oxides with Microporous Walls.
Jiao F and Bruce PG
Angewandte Chemie (International Edition in English), 43(44), 5958-5961 (2004)
M Tenne et al.
Journal of neurochemistry, 44(5), 1373-1377 (1985-05-01)
Deamination of n-octylamine and n-decylamine has been studied in various tissues using a new bioluminescence technique. Selectivity of n-octylamine and n-decylamine as substrates for monoamine oxidase (MAO) A or B has been determined using both clorgyline and (-)-deprenyl inhibition curves
Rebecca A Bader et al.
Biomacromolecules, 12(2), 314-320 (2011-01-12)
Despite improvements relative to unmodified counterparts, poly(ethylene glycol) (PEG) conjugation may not be the ideal solution for improving circulatory stability of current nanoparticle carriers or free drugs. Polysialic acid (PSA), a natural polymer for which the body possesses no receptors
Aneta D Petelska et al.
Colloids and surfaces. B, Biointerfaces, 82(2), 340-344 (2010-10-05)
Monolayers of phosphatidylcholine, fatty acid and amine and binary mixtures phosphatidylcholine-fatty acid or phosphatidylcholine-amine were investigated at the air/water interface. Phosphatidylcholine (lecithin, PC), stearic acid (SA), palmitic acid (PA), decanoic acid (DA) and decylamine (DE) were used to the experiment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service