Skip to Content
Merck
All Photos(3)

Documents

S8502

Sigma-Aldrich

(−)-Scopolamine methyl bromide

≥98% (HPLC), powder

Synonym(s):

Hyoscine methyl bromide, Methscopolamine bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24NO4 · Br
CAS Number:
Molecular Weight:
398.29
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]25/D −25°, c = 5 in H2O(lit.)

color

white

solubility

H2O: 50 mg/mL

SMILES string

[Br-].[H][C@]12C[C@@H](C[C@]([H])([C@@]3([H])O[C@@]13[H])[N+]2(C)C)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C18H24NO4.BrH/c1-19(2)14-8-12(9-15(19)17-16(14)23-17)22-18(21)13(10-20)11-6-4-3-5-7-11;/h3-7,12-17,20H,8-10H2,1-2H3;1H/q+1;/p-1/t12-,13-,14-,15+,16-,17+;/m1./s1

InChI key

CXYRUNPLKGGUJF-RAFJPFSSSA-M

Looking for similar products? Visit Product Comparison Guide

Application

(−)-Scopolamine methyl bromide was administered to rats to inhibit the peripheral cholinergic effects induced by pilocarpine.10,11

Biochem/physiol Actions

Competitive muscarinic acetylcholine receptor antagonist.
Scopolamine is an anti-muscarinic and cholinolytic alkaloid that inhibits parasympathetic-cholinergic system.7 The central effects include hallucinations, disorientation, restlessness and euphoria.8 Scopolamine disturb the stimulus detection performance in rats by interfering with the reticular cholinergic pathways.9

Caution

Shelf-life of the powder is at least five years when stored at room temperature.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas H Varvel et al.
Neuropharmacology, 224, 109356-109356 (2022-12-03)
Cognitive comorbidities can substantially reduce quality of life in people with epilepsy. Inflammation is a component of all chronic diseases including epilepsy, as well as acute events like status epilepticus (SE). Neuroinflammation is the consequence of several broad signaling cascades
Zuzanna Setkowicz et al.
Epilepsia, 44(10), 1267-1273 (2003-09-27)
To determine whether brains irradiated at different stages of prenatal development also have different postnatal susceptibility to seizures evoked by pilocarpine. Pregnant Wistar rats were exposed to a single 1.0-Gy dose of gamma rays on gestation days 13, 15, 17
Philipp Singer et al.
Pharmacology, biochemistry, and behavior, 101(1), 107-114 (2012-01-03)
Prepulse inhibition (PPI) of the acoustic startle reflex refers to the reduction of the startle response to an intense acoustic pulse stimulus when it is shortly preceded by a weak non-startling prepulse stimulus and provides a cross-species measure of sensory-motor
Misty M Strain et al.
iScience, 27(3), 109137-109137 (2024-02-29)
Cardiovagal neurons (CVNs) innervate cardiac ganglia through the vagus nerve to control cardiac function. Although the cardioinhibitory role of CVNs in nucleus ambiguus (CVNNA) is well established, the nature and functionality of CVNs in dorsal motor nucleus of the vagus
Byron K Y Bitanihirwe et al.
Behavioural brain research, 217(1), 178-187 (2010-10-27)
Deterioration in attention and related processes is an early sign in schizophrenia predictive of disease development. Amongst the various translational paradigms for assessing attention in rodents, it is not known if they are equivalent in detecting individual differences. Answers here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service