Skip to Content
Merck
All Photos(1)

Key Documents

S3567

Sigma-Aldrich

SB 415286

≥98% (HPLC)

Synonym(s):

3-[(3-Chloro-4-hydroxyphenyl)­amino]-4-(2-nitrophenyl)-1H-pyrrol-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N3O5Cl
CAS Number:
Molecular Weight:
359.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

color

yellow to orange

solubility

DMSO: 5 mg/mL, clear
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

Oc1ccc(NC2=C(C(=O)NC2=O)c3ccccc3[N+]([O-])=O)cc1Cl

InChI

1S/C16H10ClN3O5/c17-10-7-8(5-6-12(10)21)18-14-13(15(22)19-16(14)23)9-3-1-2-4-11(9)20(24)25/h1-7,21H,(H2,18,19,22,23)

InChI key

PQCXVIPXISBFPN-UHFFFAOYSA-N

Gene Information

Application

SB 415286 was used to treat neuroblastoma cells and study the effect of GSK-3 inhibition on cell proliferation.

Biochem/physiol Actions

Glycogen synthase kinase-3 (GSK-3) inhibitor.
SB 415286 is a small molecule inhibitor of GSK-3 in muscle and fat cells. SB 415286 induces activation of glycogen synthase and regulates the transport glucose. SB 415286 reduces the systemic inflammation induced by endotoxic shock in rat model of acute colitis. It increases the axonal growth and promotes the recovery of injured adult CNS neurons. SB 415289 is implicated in inducing chromosome instability when used as therapeutic agents.

Features and Benefits

This compound is featured on the GSK-3 and PKB/Akt pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from GlaxoSmithKline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrina MacAulay et al.
European journal of biochemistry, 270(18), 3829-3838 (2003-09-03)
Glycogen synthase kinase 3 (GSK3) is inactivated by insulin and lithium and, like insulin, Li also activates glycogen synthase (GS) via inhibition of GSK3. Li also mimics insulin's ability to stimulate glucose transport (GT), an observation that has led to
Hye Ree Yoon et al.
International journal of molecular sciences, 21(18) (2020-09-23)
Dual-specificity tyrosine phosphorylation-regulated kinase 1A (DYRK1A) is a protein kinase with diverse functions in cell regulation. Abnormal expression and activity of DYRK1A contribute to numerous human malignancies, Down syndrome, and Alzheimer's disease. Notably, DYRK1A has been proposed as a potential
Daniela Hulcová et al.
Molecules (Basel, Switzerland), 23(4) (2018-03-22)
Glycogen synthase kinase-3β (GSK-3β) is a multifunctional serine/threonine protein kinase that was originally identified as an enzyme involved in the control of glycogen metabolism. It plays a key role in diverse physiological processes including metabolism, the cell cycle, and gene
Jorge-Tonatiuh Ayala-Sumuano et al.
Scientific reports, 1, 178-178 (2012-02-23)
Adipogenesis is regulated by a complex cascade of transcriptional factors, but little is known about the early events that regulate the adipogenic program. Here, we report the role of the srebf1a gene in the differentiation of fibroblastic 3T3-F442A cells. We
Brendan J R Whittle et al.
British journal of pharmacology, 147(5), 575-582 (2005-11-30)
The effects of the inhibitors of glycogen synthase kinase-3beta (GSK-3beta), TDZD-8 and SB 415286, which can substantially reduce the systemic inflammation associated with endotoxic shock in vivo, have now been investigated on the acute colitis provoked by trinitrobenzene sulphonic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service