Skip to Content
Merck
All Photos(1)

Key Documents

SML0084

Sigma-Aldrich

IM-12

≥98% (HPLC)

Synonym(s):

3-(4-Fluorophenylethylamino)-1-methyl-4-(2-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20FN3O2
CAS Number:
Molecular Weight:
377.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow-orange

solubility

DMSO: ≥9 mg/mL

storage temp.

2-8°C

SMILES string

CN1C(=O)C(NCCc2ccc(F)cc2)=C(C1=O)c3c(C)[nH]c4ccccc34

InChI

1S/C22H20FN3O2/c1-13-18(16-5-3-4-6-17(16)25-13)19-20(22(28)26(2)21(19)27)24-12-11-14-7-9-15(23)10-8-14/h3-10,24-25H,11-12H2,1-2H3

InChI key

ZKJAZFUFPPSFCO-UHFFFAOYSA-N

Biochem/physiol Actions

GSK-3β inhibitor
IM-12 is a cell-permeable indolylmaleimide that acts as a Glycogen synthase kinase 3β (GSK-3β) inhibitor, activating downstream components of canonical Wnt signaling. IM-12 significantly increased β-catenin levels. When used to treat human neural progenitor cells, IM-12 promoted neuronal differentiation resulting in an increase of neuronal cells. It has an IC50 of 53 nM in an in vitro binding assay, compared with 92 nM for SB-216763 in the same assay condition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quanxin Wu et al.
Molecular medicine reports, 23(5) (2021-03-25)
Hepatocellular carcinoma (HCC) is a malignant tumor located in the liver. Secreted frizzled‑related protein 4 (sFRP‑4) is associated with cancer occurrence, but the relationship between sFRP‑4 and HCC is not completely understood. The present study aimed to investigate the role and
Johannes Menzel-Severing et al.
Scientific reports, 8(1), 10268-10268 (2018-07-08)
Understanding transcription factor (TF) regulation of limbal epithelial stem/progenitor cells (LEPCs) may aid in using non-ocular cells to regenerate the corneal surface. This study aimed to identify and characterize TF genes expressed specifically in LEPCs isolated from human donor eyes
Xianyan Chen et al.
Frontiers in pharmacology, 10, 1528-1528 (2020-02-11)
Dihydrotestosterone (DHT) is the most potent androgen that regulates hair cycling. Hair cycling involves cross-talk between the androgen and Wnt/β-catenin pathways. However, how DHT regulates hair follicle (HF) growth through the Wnt/β-catenin pathway has not been well investigated. This study
Yongli Shan et al.
Nature communications, 8(1), 672-672 (2017-09-25)
Polycomb repressive complex 2 and the epigenetic mark that it deposits, H3K27me3, are evolutionarily conserved and play critical roles in development and cancer. However, their roles in cell fate decisions in early embryonic development remain poorly understood. Here we report
Marta N Shahbazi et al.
Nature, 552(7684), 239-243 (2017-12-01)
The foundations of mammalian development lie in a cluster of embryonic epiblast stem cells. In response to extracellular matrix signalling, these cells undergo epithelialization and create an apical surface in contact with a cavity, a fundamental event for all subsequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service