RAB0740
Human THBS1 / Thrombospondin-1 ELISA Kit
for serum, plasma, cell culture supernatants and urine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 ng/mL
standard curve range: 8.19-2000 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... THBS1(7057)
General description
The antibody pair provided in this kit recognizes Human Thrombospondin 1.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Norifumi Urao et al.
Human molecular genetics, 26(24), 4951-4960 (2017-12-06)
The purpose of this study was to determine whether thrombospondin (TSP)-1 promotes macrophage activity and disease progression in dysferlinopathy. First, we found that levels of TSP-1 are elevated in blood of non-ambulant dysferlinopathy patients compared with ambulant patients and healthy
Laetitia Cicchelero et al.
Cancer letters, 400, 205-218 (2016-10-22)
The immunological, anti-angiogenic and clinical effects of metronomic cyclophosphamide and 3 consecutive intratumoral interleukin (IL)-12 gene therapy (electrogene therapy (EGT)) treatments were evaluated in 6 dogs with spontaneous cancer. In all dogs, a decrease in peripheral leukocytes 2 days after
Ruby Maharjan et al.
Pharmaceutics, 11(7) (2019-07-25)
Metronomic chemotherapy (MCT) is defined as the frequent administration of low-dose chemotherapeutics, without long drug-free periods, with the exertion of antitumor activity exclusively through anti-angiogenic mechanisms. In this study, we have developed an orally available formulation of pemetrexed (PMX) for
Dong Hyun Kim et al.
Scientific reports, 8(1), 354-354 (2018-01-13)
Alzheimer's disease (AD) is an incurable neurodegenerative disease characterised clinically by learning and memory impairments. Amyloid beta (Aβ) peptide-induced synaptic dysfunction is a pathological process associated with early-stage AD. Here, we show that paracrine action of human umbilical cord blood-derived-mesenchymal
Hailing Xia et al.
Environmental research, 193, 110467-110467 (2020-11-17)
Malignant mesothelioma (MM) is an aggressive cancer linked to asbestos exposure. Its poor prognosis makes early diagnosis extremely important, which would provide an opportunity for early treatment and potentially changing outcomes. This study aimed to explore the underlying mechanisms of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service