Skip to Content
Merck
All Photos(1)

Documents

SRP3172

Sigma-Aldrich

Thrombomodulin human

recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

BDCA-3, CD141, THBD, TM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

51.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... THBD(7056)

General description

Thrombomodulin is an integral membrane protein that spans a length of 559 residues and contains five structural domains. The hydrophobic amino terminal domain is made up of 223 amino acid residues. The adjacent domain contains six EGF type B repeats spanning a length of 236 residues. This is followed by a 34 amino acid-long Ser/Thr-rich domain, which may be involved in 0-linked glycosylation. The next domain is a putative membrane-spanning domain of 23 residues, followed by a carboxyl-terminal cytosolic tail of 38 residues. The gene encoding this protein lacks introns. Recombinant soluble thrombomodulin is a 491 amino acid glycoprotein containing the extracellular domain of thrombomodulin.

Application

Thrombomodulin human has been used to bind to anti-TM for peptide cleavage.

Biochem/physiol Actions

Thrombomodulin (TM) is an endothelial cell expressed transmembrane glycoprotein that can form a complex with the coagulation factor, thrombin. The thrombomodulin/thrombin complex converts protein C to its activated form, protein Ca, which in turn proteolytically cleaves and deactivates factor Va and factor VIIIa, two essential components of the coagulation mechanism. This inactivation reduces the generation of additional thrombin and thereby effectively prevents continued coagulation. Reduced levels of thrombomodulin can correlate with the pathogenesis of certain cardiovascular diseases, such as atherosclerosis and thrombosis. However, the serum levels of the truncated circulating form of thrombomodulin are typically elevated during inflammation and in the presence of various inflammatory related diseases.

Sequence

APAEPQPGGS QCVEHDCFAL YPGPATFLNA SQICDGLRGH LMTVRSSVAA DVISLLLNGD GGVGRRRLWI GLQLPPGCGD PKRLGPLRGF QWVTGDNNTS YSRWARLDLN GAPLCGPLCV AVSAAEATVP SEPIWEEQQC EVKADGFLCE FHFPATCRPL AVEPGAAAAA VSITYGTPFA ARGADFQALP VGSSAAVAPL GLQLMCTAPP GAVQGHWARE APGAWDCSVE NGGCEHACNA IPGAPRCQCP AGAALQADGR SCTASATQSC NDLCEHFCVP NPDQPGSYSC MCETGYRLAA DQHRCEDVDD CILEPSPCPQ RCVNTQGGFE CHCYPNYDLV DGECVEPVDP CFRANCEYQC QPLNQTSYLC VCAEGFAPIP HEPHRCQMFC NQTACPADCD PNTQASCECP EGYILDDGFI CTDIDECENG GFCSGVCHNL PGTFECICGP DSALARHIGT DCDSGKVDGG DSGSGEPPPS PTPGSTLTPP A

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enrico Di Cera
Molecular aspects of medicine, 29(4), 203-254 (2008-03-11)
Thrombin is a Na+-activated, allosteric serine protease that plays opposing functional roles in blood coagulation. Binding of Na+ is the major driving force behind the procoagulant, prothrombotic and signaling functions of the enzyme, but is dispensable for cleavage of the
Neeraj Maheshwari et al.
PloS one, 11(3), e0150315-e0150315 (2016-03-15)
Streptokinase (SK) is a potent clot dissolver but lacks fibrin clot specificity as it activates human plasminogen (HPG) into human plasmin (HPN) throughout the system leading to increased risk of bleeding. Another major drawback associated with all thrombolytics, including tissue
Libing Jiang et al.
BMJ open, 4(12), e006770-e006770 (2014-12-24)
Sepsis is the leading cause of mortality in non-cardiological critically ill patients. There are as many as 20 million cases of sepsis annually worldwide, with a mortality rate of around 35%. It has been reported that the dysregulation of haemostatic
Exploring traditional and nontraditional roles for thrombomodulin
Loghmani H and Conway EM
Blood, 132(2), 148-158 (2018)
F Peyvandi et al.
Journal of thrombosis and haemostasis : JTH, 14(2), 340-345 (2015-11-29)
ESSENTIALS: The differential diagnosis among thrombotic microangiopathies (TMAs) is challenging. We studied a case of TMA with neurologic symptoms, no renal impairment and normal ADAMTS-13 levels. Two novel mutations in complement factor I and thrombomodulin genes were identified. Complement-regulator genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service