Skip to Content
Merck
All Photos(1)

Key Documents

R5903

Sigma-Aldrich

Anti-Rab11 antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

western blot: 6.6 μg/mL using HeLa cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB11A(8766)
mouse ... Rab11a(53869)
rat ... Rab11a(81830)

General description

Rab11 (Ras-related protein) has three subtypes -Rab11a, Rab11b, and Rab25. Rab11a (Ras-related protein Rab-11A) belongs to the ras-like small GTP-binding protein superfamily. It is found in nonpolarized cells like fibroblasts and erythroleukemia cells. It is located on human chromosome 15q21.3-q22.31 and on mouse chromosome 9. Rab11a is expressed abundantly.

Immunogen

synthetic peptide corresponding to amino acid residues 1-17 from human Rab11. This sequence is completely conserved in human, mouse, and rat and 94% conserved in C. elegans and amphibians.

Application

Anti-Rab11 has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Rab11a (Ras-related protein Rab-11A) participates in the modulation of basolateral to apical transcytosis. It controls TRPV5/6 (Ca2+-selective members) channel trafficking. It is essential for the production of bile canalicular in WIF-B9 cells (rat hepatoma/human fibroblast hybrid cells).

Physical form

Solution in phosphate buffered saline containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O-fucosylation of the notch ligand mDLL1 by POFUT1 is dispensable for ligand function
Muller J, et al.
PLoS ONE, 9(2) (2014)
Neutrophil elastase and proteinase 3 trafficking routes in myelomonocytic cells
Kallquist L, et al.
Experimental Cell Research, 316(19), 3182-3196 (2010)
Human rab11a: transcription, chromosome mapping and effect on the expression levels of host GTP-binding proteins
Gromov PS, et al.
Febs Letters, 429(3), 359-364 (1998)
The Rab11a GTPase controls Toll-like receptor 4-induced activation of interferon regulatory factor-3 on phagosomes
Husebye H, et al.
Immunity, 33(4), 583-596 (2010)
Rab11a and myosin Vb are required for bile canalicular formation in WIF-B9 cells
Wakabayashi Y, et al.
Proceedings of the National Academy of Sciences of the USA, 15087-92, 15087-15092 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service