DCAS9PROT
dCas9-3XFLAG™-Biotin Protein
from Streptococcus pyogenes with D10A and H840A mutations, recombinant, expressed in E. coli, 1X NLS
Synonym(s):
Cas9 D10A H840A, Dead Cas9, dCas9, catalytically inactive Cas9
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.51
Recommended Products
recombinant
expressed in E. coli
Quality Level
Assay
≥90% (SDS-PAGE)
form
lyophilized powder
packaging
pkg of 1 kit (3 components)
shipped in
ambient
storage temp.
−20°C
Related Categories
General description
Recombinant dCas9-3X FLAG-Biotin protein from Streptococcus pyogenes (~165 KD) is a ready-to-use reagent for genome engineering and genomic DNA detection, isolation and imaging experiments. When combined with target-specific guide RNAs, dead Streptococcus pyogenes Cas9 protein acts as a DNA targeting protein suitable for transfection of cell cultures, microinjection, DNA pull-down, chromatin enrichment, microbial identification and in situ DNA imaging. At the C terminus, the protein is tagged with a SV40 large T antigen nuclear localization signal (NLS) for nuclear import and both a 3XFLAG epitope and biotin for anti-FLAG®-based or streptavidin-based experiments.
Application
Functional Genomics/Target Validation/Genome Editing/Molecular Biology/Synthetic Biology/DNA detection/isolation/DNA imaging
Features and Benefits
- Highly specific
- Suitable for genomic DNA detection/isolation
- Ready-to-inject/transfect
Packaging
- pkg of 50 μg (≥ 300 pmol)
- pkg of 250 μg (≥ 1500pmol)
Components
Each kit consists of:
- one vial of dCas9 recombinant protein
- one vial containing 1 mL of 1× dilution buffer
- one vial containing 1 mL of Nuclease free water with glycerol
Principle
CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be rendered inactive (dCas9) with mutations to the two protein domains, RuvC and HnH (D10A and H840A respectively), which are responsible for nuclease activity. The protein can then be programmed with a gRNA and directed to bind at a desired sequence of DNA.
Reconstitution
Lyophilized dCas9 protein should be resuspended in the Reconstitution solution provided to desired concentration. Gently tap tube to completely dissolve lyophilized powder, incubate for 10 minutes on ice, and spin tube to bring material to bottom of tube.
Other Notes
Use our CRISPR Selection Tool to order gRNA
Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins
Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins
Legal Information
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Matthew D Newton et al.
Methods in molecular biology (Clifton, N.J.), 2478, 349-378 (2022-09-06)
The discovery of CRISPR/Cas9 as an easily programmable endonuclease heralds a new era of genetic manipulation. With this comes the prospect of novel gene therapy approaches, and the potential to cure previously untreatable genetic diseases. However, reports of spurious off-target
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service