Skip to Content
Merck
All Photos(1)

Documents

R4505

Sigma-Aldrich

Rabbit Serum

USDA approved, sterile-filtered, suitable for cell culture

Synonym(s):

Serum from rabbit, lapine sera, lapine serum, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

rabbit

sterility

sterile-filtered

quality

USDA approved

composition

hemoglobin (tested)

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested
mycoplasma, tested

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Rabbit serum is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.
Rabbit Serum is used for the following applications:
  • sampling for bacteriological cultures and culture conditions
  • as a supplement in BSK-S medium in a study to evaluate the infectivity and pathogenicity of BL206 and B515 grown in BSK medium containing this product vs the medium containing Sigma rabbit serum R7136
  • light microscopy, immunocytochemistry and immunohistochemistry.
  • immunoelectron microscopy

Analysis Note

Endotoxin tested

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria A Sinetova et al.
Biochimica et biophysica acta, 1817(8), 1248-1255 (2012-06-20)
The distribution of the luminal carbonic anhydrase Cah3 associated with thylakoid membranes in the chloroplast and pyrenoid was studied in wild-type cells of Chlamydomonas reinhardtii and in its cia3 mutant deficient in the activity of the Cah3 protein. In addition
Märit Pringle et al.
Veterinary microbiology, 139(3-4), 279-283 (2009-06-27)
Various ear lesions, often caused by ear biting, are common in pigs. Some herds have a high frequency of ear necrosis, a syndrome characterized by necrotic lesions along the rim of the pinna, often bilateral and sometimes resulting in loss
Guiqing Wang et al.
Infection and immunity, 72(11), 6702-6706 (2004-10-27)
The effects of variations in Barbour-Stoenner-Kelly (BSK) medium on the infectivity and pathogenicity of Borrelia burgdorferi clinical isolates were assessed by retrospective and prospective studies using a murine model of Lyme borreliosis. Thirty of 35 (86%) mice infected with any
Andrei E Samoilov et al.
International journal of environmental research and public health, 17(12) (2020-06-18)
This article describes a lethal case of leptospirosis that occurred in Southern Russia. The Leptospira strain was isolated and characterized using a microscopic agglutination test, MALDI-TOF mass spectrometry, targeted PCR, and high-throughput sequencing. We show that molecular and mass-spectrometry methods
Guillaume Carissimo et al.
BMC genomics, 19(1), 526-526 (2018-07-11)
Anopheles mosquitoes are efficient vectors of human malaria, but it is unknown why they do not transmit viruses as well as Aedes and Culex mosquitoes. The only arbovirus known to be consistently transmitted by Anopheles mosquitoes is O'nyong nyong virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service