Skip to Content
Merck
All Photos(2)

Key Documents

PHG0008

SAFC

MES sodium salt

Pharma Manufacturing

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
Beilstein:
3765682
EC Number:
MDL number:
UNSPSC Code:
12161700

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

useful pH range

5.5-6.7

pKa 

6.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

MES Sodium is a biological buffer often referred to as a “Good′s” buffer. The pKa of MES is 5.96 which makes MES an ideal candidate for cell culture media and protein based buffer formulations to maintain a stable environment in solution. MES Sodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

MES Sodium is used in cell culture media, biopharmaceutical buffer formulations (both upstream and downstream) and diagnostic reagents. MES based buffers are used in purification bioprocesses of antibodies, peptides, proteins and blood components.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Burnham et al.
Journal of virology, 89(8), 4575-4587 (2015-02-13)
Influenza A and B viruses are human pathogens that are regarded to cause almost equally significant disease burdens. Neuraminidase (NA) inhibitors (NAIs) are the only class of drugs available to treat influenza A and B virus infections, so the development
Elena Tenenbaum et al.
The Analyst, 140(22), 7726-7733 (2015-10-13)
In this work we present a label-free optical biosensor for rapid bacteria detection using a novel peptide-mimetic compound, as the recognition element. The biosensor design is based on an oxidized porous silicon (PSiO2) nanostructure used as the optical transducer, functionalized
Deirdre A Costello et al.
Journal of virology, 89(1), 350-360 (2014-10-17)
Influenza A virus strains adapt to achieve successful entry into host species. Entry is mediated by the viral membrane protein hemagglutinin (HA), which triggers membrane fusion and genome release under acidic conditions in the endosome. In addition to changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service