Skip to Content
Merck
All Photos(2)

Key Documents

P9689

Sigma-Aldrich

Propentofylline

solid

Synonym(s):

3,7-Dihydro-3-methyl-1-(5-oxohexyl)-7-propyl-1H-purine-2,6-dione, 3-Methyl-1-(5-oxohexyl)-7-propylxanthine, HWA 285

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N4O3
CAS Number:
Molecular Weight:
306.36
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: 12.4 mg/mL

storage temp.

−20°C

SMILES string

CCCn1cnc2N(C)C(=O)N(CCCCC(C)=O)C(=O)c12

InChI

1S/C15H22N4O3/c1-4-8-18-10-16-13-12(18)14(21)19(15(22)17(13)3)9-6-5-7-11(2)20/h10H,4-9H2,1-3H3

InChI key

RBQOQRRFDPXAGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

c-AMP phosphodiesterase inhibitor; nerve growth factor stimulator; adenosine transport inhibitor; non-selective adenosine receptor antagonist.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rother et al.
Dementia and geriatric cognitive disorders, 9 Suppl 1, 36-43 (1998-08-26)
Propentofylline, a neuroprotective glial cell modulator, has been shown in preclinical studies to address some of the common pathological processes of Alzheimer's disease (AD) and vascular dementia (VaD), including glial cell activation and increased production of cytokines, free radicals, and
B Kittner et al.
Annals of the New York Academy of Sciences, 826, 307-316 (1997-11-05)
The mode of action of propentofylline (a xanthine derivative) suggested that it would have beneficial effects in patients with Alzheimer's disease or vascular dementia. In four double-blind, placebo-controlled, randomized studies, 901 patients with mild to moderate Alzheimer's disease and 359
R Mielke et al.
Alzheimer disease and associated disorders, 12 Suppl 2, S29-S35 (1998-10-13)
Pathophysiologic processes common to both vascular (multi-infarct) dementia and dementia of the Alzheimer type may include microglial activation with resultant generation of inflammatory cytokines and neurotoxic free radicals, decreased secretion of nerve growth factor by astrocytes, excess release of glutamate
Y S Gwak et al.
Neuroscience, 161(3), 895-903 (2009-04-01)
In this study, we evaluated whether astrocytic and microglial activation mediates below-level neuropathic pain following spinal cord injury. Male Sprague-Dawley (225-250 g) rats were given low thoracic (T13) spinal transverse hemisection and behavioral, electrophysiological and immunohistochemical methods were used to
C Peralta et al.
Hepatology (Baltimore, Md.), 29(1), 126-132 (1998-12-24)
This study aims to determine if the protective role of adenosine in liver ischemic preconditioning is mediated by the activation of adenosine receptors and to ascertain which of these receptors is implicated in the process. Administration of adenosine A1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service