Skip to Content
Merck
All Photos(1)

Documents

P3296

Millipore

Protein G Sepharose, Fast Flow

recombinant, expressed in E. coli, aqueous ethanol suspension

Synonym(s):

Protein G-Agarose, Fast Flow from Streptococcus sp.

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

recombinant

expressed in E. coli

form

aqueous ethanol suspension

analyte chemical class(es)

proteins (Immunoglobulins of various mammalian species)

extent of labeling

~2 mg per mL

technique(s)

affinity chromatography: suitable

matrix

Sepharose 4B Fast Flow

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protein G is an immunoglobulin (IgG)-binding bacterial cell wall protein isolated from group G streptococcal strain, G-148. This protein can be extracted from the cells by papain digestion and purified by the sequential use of ion-exchange chromatography on DEAE-Sephadex, affinity chromatography on Sepharose-coupled human IgG, and gel chromatography on Sephadex G-200. The binding between protein G and various polyclonal and monoclonal IgG is basically pH dependent between 2.8 and 10, with the strongest binding at pH 4 and 5, and weakest at pH 10. It acts as a powerful reagent for the detection of IgG.
Protein G is an immunoglobulin (IgG)-binding bacterial cell wall protein isolated from group G streptococcal strain, G-148. This protein can be extracted from the cells by papain digestion and purified by the sequential use of ion-exchange chromatography on DEAE-Sephadex, affinity chromatography on Sepharose-coupled human IgG, and gel chromatography on Sephadex G-200. The binding between protein G and various polyclonal and monoclonal IgG is basically pH dependent between 2.8 and 10, with the strongest binding at pH 4 and 5, and weakest at pH 10. It acts as a powerful reagent for the detection of IgG.

P3296-5Ml′s updated product number is GE17-0618-01

Application

Protein G-Sepharose is used in affinity chromatography, protein chromatography, antibody purification and characterization, immunoaffinity matrices, protein A, G and L resins, protein interaction, and purification and detection. Protein G-Sepharose has been used to develop a strategy to confirm the presence of anti-erythropoietin neutralizing antibodies in human serum as well as to compare methods for depletion of albumin and IgG from equine serum.

Physical form

Suspension in 20% ethanol

Preparation Note

Prepared with recombinant streptococcal Protein G from which the albumin-binding region has been genetically deleted

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Yang et al.
Oncogene, 26(1), 91-101 (2006-06-27)
The t(8;21) chromosomal translocation that generates the fusion oncoprotein RUNX1-ETO predominates in leukemia patients of the French-American-British (FAB) class M2 subtype. The oncoprotein has the capacity to promote expansion of hematopoietic stem/progenitor cells and induces leukemia in association with other
Cristina Hidalgo-Carcedo et al.
Nature cell biology, 13(1), 49-58 (2010-12-21)
Collective cell migration occurs in a range of contexts: cancer cells frequently invade in cohorts while retaining cell-cell junctions. Here we show that collective invasion by cancer cells depends on decreasing actomyosin contractility at sites of cell-cell contact. When actomyosin
B Akerström et al.
The Journal of biological chemistry, 261(22), 10240-10247 (1986-08-05)
Protein G, an IgG-binding molecule, was prepared from the cell walls of a group G streptococcal strain, G-148. The protein could be extracted from the cells by papain digestion and purified by the sequential use of ion-exchange chromatography on DEAE-Sephadex
Yi-Jye Chern et al.
Cell death & disease, 10(7), 504-504 (2019-06-28)
Therapy-refractory disease is one of the main contributors of treatment failure in cancer. In colorectal cancer (CRC), SPARC can function as a sensitizer to conventional chemotherapy by enhancing apoptosis by interfering with the activity of Bcl-2. Here, we examine a
B Akerström et al.
Journal of immunology (Baltimore, Md. : 1950), 135(4), 2589-2592 (1985-10-01)
Protein G is an immunoglobulin (IgG)-binding bacterial cell wall protein recently isolated from group G streptococci. We have investigated the avidity of protein G for various monoclonal and polyclonal Ig of the IgG class, and compared it with the binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service