GE17-1440-02
Ficoll® Paque Plus
Cytiva 17-1440-02, pack of 6 × 100 mL
Synonym(s):
Ficoll density gradient medium, cell culture media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352201
NACRES:
NA.25
Recommended Products
form
liquid
packaging
pack of 6 × 100 mL
manufacturer/tradename
Cytiva 17-1440-02
color
Colorless to slightly yellow
pH range
5.5-7.5
Related Categories
General description
Ficoll-Paque Plus is a recognized standard in laboratories worldwide for the isolation of human lymphocytes for in vitro studies.
Ficoll-Paque PLUS is a ready to use, sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.
Ficoll-Paque PLUS is a ready to use, sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.
Application
For in vitro isolation of lymphocytes from human peripheral blood.
Features and Benefits
- Ready to use sterile medium for isolation of human lymphocytes in high yield from peripheral blood.
- Maintains viability and representative distribution of B and T lymphocytes.
- Low levels of endotoxin (< 0.12 EU/mL).
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Ficoll is a registered trademark of Cytiva
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amalesh Nanda et al.
Annals of laboratory medicine, 40(5), 390-397 (2020-04-21)
The effect of the interplay among inflammation, angiogenesis, extracellular matrix (ECM) degradation, and oxidative stress (OS) on the pathogenesis of endometriosis remains unclear. Previously, we demonstrated the role of OS in endometriosis. Here, we performed a comprehensive investigation of several
Mehdi Hassanpour et al.
Stem cell research & therapy, 11(1), 139-139 (2020-03-29)
To date, many attempts are employed to increase the regenerative potential of stem cells. In this study, we evaluated the hypothesis of whether an autophagy modulation could alter differentiation potency of CD146+ cells into mature pericyte, endothelial, and cardiomyocyte lineage.
Matthew A Lakins et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(15), 4154-4167 (2020-04-30)
With the increased prevalence in checkpoint therapy resistance, there remains a significant unmet need for additional therapies for patients with relapsing or refractory cancer. We have developed FS222, a bispecific tetravalent antibody targeting CD137 and PD-L1, to induce T-cell activation
Stanislav Pantelyushin et al.
Cancers, 13(4) (2021-03-07)
Rodent cancer models have limitations in predicting efficacy, tolerability and accompanying biomarkers of ICIs in humans. Companion dogs suffering from neoplastic diseases have gained attention as a highly relevant translational disease model. Despite successful reports of PD-1/PD-L1 blockade in dogs
Jingyong Li et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(3), 446-458 (2019-01-18)
Objective- Inflammation occurs during the progression of abdominal aortic aneurysm (AAA). IL (interleukin)-33 is a pleiotropic cytokine with multiple immunomodulatory effects, yet its role in AAA remains unknown. Approach and Results- Immunoblot, immunohistochemistry, and immunofluorescent staining revealed increased IL-33 expression
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service