Skip to Content
Merck
All Photos(1)

Key Documents

P2922

Sigma-Aldrich

Endoproteinase Glu-C from Staphylococcus aureus V8

Type XVII-B, lyophilized powder, 500-1,000 units/mg solid

Synonym(s):

V8 Protease

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

type

Type XVII-B

Quality Level

form

lyophilized powder

specific activity

500-1,000 units/mg solid

mol wt

29 kDa

purified by

chromatography

shipped in

wet ice

storage temp.

−20°C

Gene Information

Staphylococcus aureus subsp. aureus MW2 ... sspA(1003044)

Looking for similar products? Visit Product Comparison Guide

Application

Endoproteinase Glu-C from Staphylococcus aureus strain V8 is a serine protease used for selective cleavage of proteins for amino acid sequence determination or peptide mapping . Product P2922 has been used to linearize cyclic peptides in C. ternatea leaf extract.
Endoproteinase Glu-C from Staphylococcus aureus V8 has been used to digest reduced and alkylated cyclotides to produce linearized fragments.
It is used for selective cleavage of proteins for amino acid sequence determination or peptide mapping.

Biochem/physiol Actions

Staphylococcus strain V8 protease specifically cleaves peptide bonds on the carboxyl side of aspartic and glutamic acid residues when used in phosphate buffer. When used in ammonium bicarbonate buffer or ammonium acetate buffer cleavage is restricted to the carboxyl side of glutamic acid residues only. The enzyme exhibits maximal activity from pH 4.0 to 7.8. If hemoglobin is used as the substrate, maximal activity is at pH 4.0. The maximal activity is at pH of 7.8 when casein is the substrate.

Unit Definition

One unit will hydrolyze 1 μmole of N-t-Boc-L-glutamic acid α-phenyl ester per min at pH 7.8 at 37 °C. One unit is equivalent to ~0.004 casein digestion unit.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From the Cover: Discovery of an unusual biosynthetic origin for circular proteins in legumes
Aaron G. Poth, Michelle L. Colgrave, et al.
Proceedings of the National Academy of Sciences of the USA, 108 (2011)
Andrew Michael Frey et al.
Cell reports, 35(1), 108930-108930 (2021-04-08)
Staphylococcus aureus possesses ten extracellular proteases with mostly unknown targets in the human proteome. To assist with bacterial protease target discovery, we have applied and compared two N-terminomics methods to investigate cleavage of human serum proteins by S. aureus V8 protease
Shanshan Liu et al.
Amino acids, 48(4), 1059-1067 (2016-01-11)
Common yet often overlooked, deamidation of peptidyl asparagine (Asn or N) generates aspartic acid (Asp or D) or isoaspartic acid (isoAsp or isoD). Being a spontaneous, non-enzymatic protein post-translational modification, deamidation artifact can be easily introduced during sample preparation, especially
Yurgis Av Yomantas et al.
Microbial cell factories, 10, 64-64 (2011-08-09)
Plasmid-less, engineered Bacillus strains have several advantages over plasmid-carrier variants. Specifically, their stability and potential ecological safety make them of use in industrial applications. As a rule, however, it is necessary to incorporate many copies of a key gene into
Hélio Nitta Matsuura
Methods in molecular biology (Clifton, N.J.), 2469, 165-181 (2022-05-05)
Cyclotides are small circular peptides carrying an array of interesting biological activities and also showing interesting features for storage and bioavailability. Here, an optimized method to isolate cyclotides from two species of Psychotria, P. brachyceras and P. leiocarpa, that can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service