Skip to Content
Merck
All Photos(4)

Documents

P1851

Sigma-Aldrich

PIPES

BioPerformance Certified, suitable for cell culture

Synonym(s):

1,4-Piperazinediethanesulfonic acid, Piperazine-1,4-bis(2-ethanesulfonic acid), Piperazine-N,N′-bis(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O6S2
CAS Number:
Molecular Weight:
302.37
Beilstein:
817713
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified
for molecular biology

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and Total Aerobic Microbial Count, tested

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>300 °C (lit.)

solubility

1 M NaOH: 20 + 80 mL g, clear, colorless
soluble

application(s)

diagnostic assay manufacturing

foreign activity

DNAse, Exonuclease; NICKase, Endonuclease; RNAse; Protease, none detected

SMILES string

OS(=O)(=O)CCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O6S2/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16/h1-8H2,(H,11,12,13)(H,14,15,16)

InChI key

IHPYMWDTONKSCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Sensitive Detection of Immunoglobulin G Stability Using in Real-Time Isothermal Differential Scanning Fluorimetry: Determinants of Protein Stability for Antibody-Based Therapeutics.: The use of PIPES buffer in this study was integral to maintaining the stability of immunoglobulin G during differential scanning fluorimetry. This method is crucial for evaluating the stability of antibody-based therapeutics (Moggridge et al., 2017).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Veronique Adoue et al.
Molecular systems biology, 10, 754-754 (2014-10-19)
Most complex disease-associated genetic variants are located in non-coding regions and are therefore thought to be regulatory in nature. Association mapping of differential allelic expression (AE) is a powerful method to identify SNPs with direct cis-regulatory impact (cis-rSNPs). We used
Kalina Makowiecki et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(32), 10780-10792 (2014-08-08)
Repetitive transcranial magnetic stimulation (rTMS) is increasingly used as a treatment for neurological and psychiatric disorders. Although the induced field is focused on a target region during rTMS, adjacent areas also receive stimulation at a lower intensity and the contribution
B W Nielsen et al.
Agents and actions, 35(3-4), 170-178 (1992-03-01)
Increased osmotic pressure has been reported to cause non-cytotoxic histamine release (HR) from human basophils, as well as a potentiation of HR induced by anti-IgE. In this study, the effects of hyperosmolar Na-K-acetate (300-600 mOsm/kg H2O) on HR was studied
V D Prokopieva et al.
Alcohol and alcoholism (Oxford, Oxfordshire), 35(1), 44-48 (2000-02-24)
The effects of carnosine and related compounds on erythrocytes from alcoholics were studied. In their presence, erythrocytes showed an increased ability to resist haemolysis and showed a more normal morphology, with carnosine and N-acetyl-carnosine being the most effective compounds. These
Leticia A Montoya et al.
Analytical chemistry, 86(12), 6032-6039 (2014-05-24)
Sulfhydryl-containing compounds, including thiols and hydrogen sulfide (H2S), play important but differential roles in biological structure and function. One major challenge in separating the biological roles of thiols and H2S is developing tools to effectively separate the reactivity of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service