Skip to Content
Merck
All Photos(5)

Key Documents

T0377

Sigma-Aldrich

Tricine

≥99% (titration)

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (titration)

form

crystalline powder

useful pH range

7.4-8.8

pKa (25 °C)

8.1

mp

187 °C

solubility

water: 0.25 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tricine is the most frequently used electrophoresis buffer. It is also useful for the resuspension of cell pellets. Tricine functions as a trailing ion and aids the resolution of small proteins at lower acrylamide concentrations than in glycine-sodium dodecyl-sulfate polyacrylamide gel electrophoresis (SDS-PAGE) systems. It has a lower negative charge than glycine, which helps it migrate faster. Tricine has a high ionic strength that allows increased ion movement and less protein movement. This in turn leads to the separation of low molecular weight proteins in lower percent acrylamide gels.

Application

Buffer component for separation of low molecular weight peptides.
Tricine has been used:
  • to prevent precipitation of salts during autoclaving of Emiliania huxleyi cultures
  • as a component of buffer A for the homogenization of samples like Caenorhabditis elegans, Drosophila, and plants
  • as a component of fresh assay buffer to measure serum melatonin by radioimmunoassay

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Encyclopedia of Dairy Sciences (2011)
Assessing the applicability of Emiliania huxleyi coccolith morphology as a sea-surface salinity proxy
Fielding SR, et al.
Limnology and Oceanography, 54(5), 1475-1480 (2009)
Elzbieta Turkowska et al.
Chronobiology international, 31(1), 134-143 (2013-10-19)
The avian pineal gland, apart from the hypothalamic master clock (suprachiasmatic nuclei, SCN) and retina, functions as an independent circadian oscillator, receiving external photic cues that it translates into the rhythmical synthesis of melatonin, a biochemical signal of darkness. Functional
Yunfeng Ding et al.
Nature protocols, 8(1), 43-51 (2012-12-12)
The lipid droplet (LD) is a cell organelle that has been linked to human metabolic syndromes and that can be exploited for the development of biofuels. The isolation of LDs is crucial for carrying out morphological and biochemical studies of
Christian Nilsson et al.
Electrophoresis, 31(3), 459-464 (2010-02-02)
Totally porous lipid-based liquid crystalline nanoparticles were used as pseudostationary phase for capillary electroseparation with LIF detection of proteins at physiological conditions using unmodified cyclic olefin copolymer capillaries (Topas, 6.7 cm effective length). In the absence of nanoparticles, i.e. in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service