P0089
Anti-Prox1 antibody , Mouse monoclonal
clone 4G10, purified from hybridoma cell culture
Synonym(s):
Anti-Homeobox prospero-like protein Prox1, Anti-Prospero homeobox 1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
4G10, monoclonal
form
buffered aqueous solution
mol wt
antigen ~84 kDa
species reactivity
chicken, rat, lizard, human
concentration
~1.5 mg/mL
technique(s)
immunocytochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 2.5-5 μg/mL using paraformaldehyde fixed HepG2 cells
western blot: suitable
isotype
IgG2b
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PROX1(5629)
rat ... Prox1(305066)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service