Skip to Content
Merck
All Photos(2)

Documents

L6545

Sigma-Aldrich

Letrozole

≥98% (HPLC), powder, non-steroidal aromatase inhibitor

Synonym(s):

4,4′-(1H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N5
CAS Number:
Molecular Weight:
285.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

Letrozole, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >50 mg/mL

originator

Novartis

storage temp.

2-8°C

SMILES string

N#CC(C=C1)=CC=C1C(N2C=NC=N2)C3=CC=C(C#N)C=C3

InChI

1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H

InChI key

HPJKCIUCZWXJDR-UHFFFAOYSA-N

Gene Information

human ... CYP19A1(1588)

Looking for similar products? Visit Product Comparison Guide

Application

Letrozole has been used:
  • in organoid growth assay to determine its inhibitory capacity(48)
  • to investigate steroid receptor coactivator-1 (SRC-1) mediated endogenous estrogen regulation of hippocampal PSD-95(49)
  • to determine its effects on tumor-induced hyperalgesia(50)
  • for hormonal manipulation in rats(51)
  • to study its effects on lipocalin-2 (Lcn2)(52)
  • to determine its effects on mechanical hyperalgesia and aromatase expression(53)

Biochem/physiol Actions

Letrozole acts as an adjuvant agent and is used to treat breast cancer.
Letrozole is a third generation nonsteroidal aromatase inhibitor. It is a competitive inhibitor of the aromatase enzyme system and thus inhibits the conversion of androgens to estrogens. Letrozole inhibits the aromatase enzyme by competitively binding to the heme of the cytochrome P450 subunit of the enzyme, resulting in a reduction of estrogen biosynthesis in all tissues.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Posocco et al.
PloS one, 15(2), e0228822-e0228822 (2020-02-08)
A novel LC-MS/MS method was developed for the quantification of the new cyclin dependent kinase inhibitors (CDKIs) palbociclib and ribociclib and the aromatase inhibitor letrozole used in combinatory regimen. The proposed method is appropriate to be applied in clinical practice
Zahra Shaaban et al.
Iranian journal of basic medical sciences, 25(1), 110-120 (2022-06-04)
Despite prevalence of polycystic ovary syndrome (PCOS) among childbearing women and development of many animal models for this syndrome, information on its etiology is still scarce. The intrauterine hyperandrogenic environment may underlie changes at the level of hypothalamus, pituitary, ovary
Lijun Tan et al.
Cancers, 14(19) (2022-10-15)
Antiestrogen therapy (AET) is an alternative to cytotoxic chemotherapy for recurrent ovarian cancer, yet the often short duration of response suggests mechanisms of resistance. We previously demonstrated that tumor microenvironment interleukin-6/leukemia inhibitory factor (IL6/LIF) cytokines induce tumor cell JAK-STAT signaling
G Allevi et al.
British journal of cancer, 108(8), 1587-1592 (2013-04-13)
The objective of this study was to determine the optimal scheduling of 2.5 mg daily letrozole in neoadjuvant breast cancer patients to obtain pathological complete response (pathCR) and assess Ki-67 expression as an early predictor of response. This single institution
Natalia Lagunas et al.
Frontiers in neuroanatomy, 16, 902218-902218 (2022-07-12)
Sex steroid hormones, such as androgens and estrogens, are known to exert organizational action at perinatal periods and activational effects during adulthood on the brain and peripheral tissues. These organizational effects are essential for the establishment of biological axes responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service