Skip to Content
Merck
All Photos(2)

Key Documents

OGS1879

Sigma-Aldrich

PSF-TEF1-NH2-INTS - INVERTASE SECRETION PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

form

buffered aqueous solution

mol wt

size 6787 bp

bacteria selection

kanamycin

Origin of replication

2Micron
pUC (500 copies)

Peptide cleavage

no cleavage

Peptide tag location

N-terminal

Promoter

Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast

reporter gene

none

Secretion signal

alpha factor FL

shipped in

ambient

storage temp.

−20°C

yeast selection

uracil

General description

This plasmid is designed to express tagged proteins in yeast cells (Saccharomyces cerevisiae). The plasmid contains an auxotrophic Uracil selection expression cassette (URA3) that allows for the positive selection of yeast that are deficient in the URA3 gene (YEL021W). This is typically achieved by growing the yeast in minimal media that is reconstituted with the essential amino acids and nucleotides but excluding Uracil.

About the Peptide Tag:This plasmid contains an Invertase (Ints) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases.

Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.

Sequence

To view sequence information for this product, please visit the product page

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service