Skip to Content
Merck
All Photos(3)

Key Documents

CMC0014

Sigma-Aldrich

BL21(DE3) Chemically Competent Cells

Escherichia coli, rod shaped

Synonym(s):

Competent E. coli cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

BL21(DE3) Chemically Competent Cells, for protein expression

biological source

Escherichia coli

grade

Molecular Biology
for molecular biology

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: chemically competent
transformation efficiency: ≥1 x 107 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The BL21(DE3) Chemically Competent strains contain the phage T7 RNA polymerase gene linked to the IPTG-inducible promoter, for use with any expression plasmid containing the T7 promoter.

Genotype

F – ompT hsdSB (rB- mB-) gal dcm (DE3)

Application

The E. coli strain BL21 (DE3) has been used:

  • to express tau 2N4R isoform (aa1–441) and the truncated tau (aa1–421) using pET19b as expression vector.[1]
  • BL21(DE3)Chemically Competent Cells has been used for the transformation and expressionof protegrin-1 (PG1) peptides.[2]
  • to express 6His-TEVsite-Hs ARF1 (17-181)-Q71L in pHis2 vector.[3][1]

Features and Benefits

  • a transformation efficiency of >1 × 107 cfu/μg.
  • economical prices
  • offering superior value for everyday protein expression work.

Components

  • BL21(DE3) chemically competent cells
  • pUC 19 transformation control DNA
  • recovery medium for expression

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasaman Damestani et al.
Lasers in surgery and medicine, 48(8), 782-789 (2016-10-19)
The development and feasibility of a novel nanocrystalline yttria-stabilized-zirconia (nc-YSZ) cranial implant has been recently established. The purpose of what we now call "window to the brain (WttB)" implant (or platform), is to improve patient care by providing a technique
David B Sauer et al.
eLife, 9 (2020-09-02)
Citrate, α-ketoglutarate and succinate are TCA cycle intermediates that also play essential roles in metabolic signaling and cellular regulation. These di- and tricarboxylates are imported into the cell by the divalent anion sodium symporter (DASS) family of plasma membrane transporters
A validated antibody panel for the characterization of tau post-translational modifications.
Ebru E, et al.
Mol. Neurodegener., 12(1), 87-87 (2017)
Romana Koszagova et al.
Microbial cell factories, 17(1), 139-139 (2018-09-05)
Physiological aggregation of a recombinant enzyme into enzymatically active inclusion bodies could be an excellent strategy to obtain immobilized enzymes for industrial biotransformation processes. However, it is not convenient to recycle "gelatinous masses" of protein inclusion bodies from one reaction
Gints Kalnins et al.
Nature communications, 11(1), 388-388 (2020-01-22)
Bacterial microcompartments (BMCs) are prokaryotic organelles consisting of a protein shell and an encapsulated enzymatic core. BMCs are involved in several biochemical processes, such as choline, glycerol and ethanolamine degradation and carbon fixation. Since non-native enzymes can also be encapsulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service