Skip to Content
Merck
All Photos(1)

Key Documents

N6383

Sigma-Aldrich

Neurotensin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H121N21O20
CAS Number:
Molecular Weight:
1672.92
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H]2CCCN2C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCCCN)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]5CCC(=O)N5)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C78H121N21O20/c1-7-43(6)63(73(115)96-57(76(118)119)37-42(4)5)97-70(112)55(39-45-21-25-47(101)26-22-45)95-72(114)59-18-13-35-99(59)75(117)52(16-11-33-86-78(83)84)90-64(106)48(15-10-32-85-77(81)82)89-71(113)58-17-12-34-98(58)74(116)51(14-8-9-31-79)91-69(111)56(40-60(80)102)94-66(108)50(28-30-62(104)105)88-68(110)54(38-44-19-23-46(100)24-20-44)93-67(109)53(36-41(2)3)92-65(107)49-27-29-61(103)87-49/h19-26,41-43,48-59,63,100-101H,7-18,27-40,79H2,1-6H3,(H2,80,102)(H,87,103)(H,88,110)(H,89,113)(H,90,106)(H,91,111)(H,92,107)(H,93,109)(H,94,108)(H,95,114)(H,96,115)(H,97,112)(H,104,105)(H,118,119)(H4,81,82,85)(H4,83,84,86)/t43-,48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,63-/m0/s1

InChI key

PCJGZPGTCUMMOT-ISULXFBGSA-N

Gene Information

human ... NTS(4922)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-Leu-Tyr-Glu-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu

General description

Neurotensin (NT) is encoded by the gene mapped to human chromosome 12q21.31.[1] The encoded protein is a tridecapeptide with pGlu–Leu–Tyr–Glu–Asn–Lys–Pro–Arg–Arg–Pro–Tyr–Ile–Leu–COOH amino acid sequence.[2] NT is widely distributed in the gastrointestinal (GI) tract and central nervous system (CNS), but at high levels in hypothalamus, amygdala and nucleus accumbens as well as at low levels in peripheral blood.[1][3]

Application

Neurotensin has been used as an external calibration reagent in spectrum calibration.[4] It has also been used to evaluate the performance of online capillary electrophoresis to matrix-assisted laser desorption ionization (CE-MALDI).[5]
Neurotensin has been used in apoptosis/ cell death assay and is used in:
  • matrix-assisted laser desorption/ionisation-time of flight (MALDI-TOF) mass spectrometry analysis[6][7][8]

Biochem/physiol Actions

Neurotensin (NT) acts as an inducer for analgesia, hypothermia and hyperglycemia, and is an inhibitor of gastric motility. Plasma NT levels are high in children with Prader–Willi syndrome (PWS).[1] NT secreted from small cell lung cancer cells might have a regulatory action in this disease.[9] It has been implicated in regulation of gastric system and inflammatory processes in the lung. In addition, NT also regulates immune response by stimulating cytokine release and chemotaxis on interaction with leukocytes, peritoneal mast cells and dendritic cells.[3] NT present in gastrointestinal (GI) tract plays a vital role in various processes such as stimulation of pancreatic and biliary secretion and enhances growth of various normal tissues. It is involved downstream of Wnt/β-catenin signaling pathway and stimulates the growth of neuroendocrine tumor cells.[10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proneurotrophin-3 may induce Sortilin-dependent death in inner ear neurons.
Tauris J
The European Journal of Neuroscience, 33(4), 622-631 (2011)
MALDI TOF-TOF characterization of a light stabilizer polymer contaminant from polypropylene or polyethylene plastic test tubes.
Sachon E
Journal of Mass Spectrometry : Jms, 45(1), 43-50 (2010)
Autohydrolysis of a partially cyclized mu/nu-carrageenan and structural elucidation of the oligosaccharides by chemical analysis, NMR spectroscopy and UV-MALDI mass spectrometry
Ciancia M, et al.
ARKIVOC (Gainesville, FL, United States), 12, 319-331 (2005)
High plasma neurotensin levels in children with Prader-Willi syndrome.
Butler MG
American Journal of Medical Genetics. Part A, 167A(8), 1773-1778 (2015)
Hypothermia and intolerance to cold induced by intracisternal administration of the hypothalamic peptide neurotensin.
Nature, 262(5569), 607-609 (1976)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service