Skip to Content
Merck
All Photos(1)

Key Documents

O6262

Sigma-Aldrich

Orexin B rat, mouse

≥97% (HPLC)

Synonym(s):

Hypocretin 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C126H215N45O34S
CAS Number:
Molecular Weight:
2936.40
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Assay:
≥97% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

solubility

1% acetic acid: 1.00-1.04 mg/mL, clear, colorless
water: 1.00-1.04 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(

InChI

1S/C126H215N45O34S/c1-18-65(12)99(120(202)166-82(49-64(10)11)117(199)168-100(69(16)172)121(203)156-72(101(133)183)37-44-206-17)167-97(181)57-144-102(184)66(13)150-103(185)67(14)152-111(193)83(50-70-53-140-59-149-70)165-116(198)85(52-93(132)177)155-95(179)55-146-106(188)84(51-92(131)176)161-104(186)68(15)151-107(189)76(32-35-90(129)174)159-114(196)80(47-62(6)7)164-115(197)81(48-63(8)9)163-109(191)74(27-21-40-143-126(138)139)157-110(192)77(33-36-91(130)175)160-113(195)79(46-61(4)5)162-108(190)73(26-20-39-142-125(136)137)153-94(178)54-145-105(187)75(31-34-89(128)173)158-112(194)78(45-60(2)3)154-96(180)56-147-118(200)87-29-23-43-171(87)123(205)88-30-24-41-169(88)98(182)58-148-119(201)86-28-22-42-170(86)122(204)71(127)25-19-38-141-124(134)135/h53,59-69,71-88,99-100,172H,18-52,54-58,127H2,1-17H3,(H2,128,173)(H2,129,174)(H2,130,175)(H2,131,176)(H2,132,177)(H2,133,183)(H,140,149)(H,144,184)(H,145,187)(H,146,188)(H,147,200)(H,148,201)(H,150,185)(H,151,189)(H,152,193)(H,153,178)(H,154,180)(H,155,179)(H,156,203)(H,157,192)(H,158,194)(H,159,196)(H,160,195)(H,161,186)(H,162,190)(H,163,191)(H,164,197)(H,165,198)(H,166,202)(H,167,181)(H,168,199)(H4,134,135,141)(H4,136,137,142)(H4,138,139,143)/t65-,66-,67-,68-,69+,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,99-,100-/m0/s1

InChI key

RLVZFBVFVIDDPQ-GWQFQBPDSA-N

Gene Information

mouse ... Hcrt(15171)
rat ... Hcrt(25723)

Amino Acid Sequence

Arg-Pro-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Asn-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2

General description

Orexin A, also known as hypocretin 1 (hcrt1), is a 33 amino acid excitatory neuropeptide, localized in a small set of neurons in the perifornical area of the hypothalamus. It is synthesized from the precursor protein prepro-orexin in neurons. Orexin A binds to its G-protein-coupled receptor orexin-1 (OX1), which is also called as hcrt receptors 1 (hcrtr1).

Application

Orexin B rat, mouse has been used to test its effect on social interaction in the stress-resilient mice.

Biochem/physiol Actions

Activates OX2 receptors; both OX1 and OX2 receptors are involved in maintaining arousal; OX2 receptors appear to be inhibitory autoreceptors on orexin-containing neurons.
Orexins (hypocretins) comprise a family of hypothalamic peptides that are involved in the initiation of sleep and in feeding behavior. Orexin A stimulates food intake through activation of the OX1 receptor. Orexin B activates OX2 receptors. Both OX1 and OX2 receptors are involved in maintaining arousal. OX2 receptors appear to be inhibitory autoreceptors on orexin-containing neurons.
Orexins plays a vital role in several physiological functions including feeding, neuroendocrine regulation and arousal. In addition to this, it is also associated with the pathophysiology of narcolepsy. Orexin A, expressed in locus coeruleus (LC) area, binds to orexin-1 (OX1) receptor and regulates rapid eye movement (REM) sleep by acting on the firing rate of noradrenergic neurons. Orexin A release is regulated by suprachiasmatic nucleus (SCN).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orexin administration to mice that underwent chronic stress produces bimodal effects on emotion-related behaviors
Chung HS, et al.
Regulatory Peptides, 194, 16-22 (2014)
Lesions of the suprachiasmatic nucleus eliminate the daily rhythm of hypocretin-1 release.
Zhang S
Sleep (2004)
Hypocretin-1 modulates rapid eye movement sleep through activation of locus coeruleus neurons.
Bourgin P
The Journal of Neuroscience (2000)
Hoyoung An et al.
Journal of Alzheimer's disease : JAD, 58(1), 253-261 (2017-04-08)
Intracranial accumulation of amyloid-β (Aβ) is a characteristic finding of Alzheimer's disease (AD). It is thought to be the result of Aβ overproduction by neurons and impaired clearance by several systems, including degradation by microglia. Sleep disturbance is now considered
Xing Xiao et al.
Nature communications, 12(1), 5249-5249 (2021-09-04)
The wake-active orexin system plays a central role in the dynamic regulation of glucose homeostasis. Here we show orexin receptor type 1 and 2 are predominantly expressed in dorsal raphe nucleus-dorsal and -ventral, respectively. Serotonergic neurons in ventral median raphe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service