Skip to Content
Merck
All Photos(1)

Key Documents

N4767

Sigma-Aldrich

N-Z-Amine® EKC

Casein enzymatic hydrolysate, from bovine milk

Synonym(s):

Peptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352116
NACRES:
NA.85
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

pH

6.4-7.2(2% solution)

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

NZ Amine EKC is derived from the enzymatic digest of casein, often used in culture media. NZ Amine EKC contains a mixture of peptides and amino acids.

Features and Benefits

NZ Amine powder provides:
  • Granulated, dust-free format for safer handling and faster mixing
  • Easy scale-up using larger package sizes

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerald F Bills et al.
The Journal of antibiotics, 69(3), 141-148 (2015-10-16)
The invalidly published name Aspergillus sydowii var. mulundensis was proposed for a strain of Aspergillus that produced new echinocandin metabolites designated as the mulundocadins. Reinvestigation of this strain (Y-30462=DSMZ 5745) using phylogenetic, morphological, and metabolic data indicated that it is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service