Skip to Content
Merck
All Photos(4)

Key Documents

22090

Millipore

Casein Peptone

Enzymatic digest of casein from bovine milk, ≤3.5% chloride, suitable for biotechnology and microbiology

Synonym(s):

Peptone from casein, Casein Hydrolysate, Pancreatic hydrolysate of casein, peptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

Product Name

Casein Hydrolysate, suitable for microbiology, Nitrogen source in culture media

biological source

bovine milk

Quality Level

form

powder

composition

total nitrogen (N), ≥11%

storage condition

(sealed container)

impurities

≤3.5% chloride (expressed as NaCl)

ign. residue

≤8%

loss

≤6.0% loss on drying

pH

6.0-7.0 (25 °C)

solubility

H2O: 2%, clear, faintly yellow to brown

application(s)

food and beverages
microbiology

storage temp.

10-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Pancreatic hydrolysate of casein is manufactured by controlled enzyme hydrolysis of milk protein. It can act as a nutrient supplement in culture media because it is rich in certain types of amino acids and hence serves as an efficient nitrogen source. It also enables the production of antibiotics, toxins, bacteriocins, enzymes, etc.

Application

Casein hydrolysate has various applications, in sterility testing media or biochemical characterization as it is utilized by a wide variety of microorganisms.It can also be used in the production of certain antibiotics, toxins, bacteriocins, and enzymes.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro M D S Abrantes et al.
Journal of microbiological methods, 169, 105815-105815 (2019-12-25)
Candida albicans and streptococci are amongst the most common fungal and bacterial organisms present in the oral cavity, with a growing body of evidence implicating C. albicans in increased caries severity and in the formation of the cariogenic biofilm. However
Kacy Greenhalgh et al.
Cell reports, 27(5), 1621-1632 (2019-05-03)
By modulating the human gut microbiome, prebiotics and probiotics (combinations of which are called synbiotics) may be used to treat diseases such as colorectal cancer (CRC). Methodological limitations have prevented determining the potential combinatorial mechanisms of action of such regimens. We expanded
Losee L Ling et al.
Nature, 517(7535), 455-459 (2015-01-07)
Antibiotic resistance is spreading faster than the introduction of new compounds into clinical practice, causing a public health crisis. Most antibiotics were produced by screening soil microorganisms, but this limited resource of cultivable bacteria was overmined by the 1960s. Synthetic

Questions

1–2 of 2 Questions  
  1. What is the molecular weight?

    1 answer
    1. Product number 22090, Casein hydrolysate, is the milk protein casein which has been treated to break the protein down into smaller peptides and amino groups.  Since it contains many different sized molecules, the product will not have a molecular weight.

      Helpful?

  2. Hello, is this product food safe?

    1 answer
    1. This product is not considered food safe. Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service