Skip to Content
Merck
All Photos(1)

Key Documents

MHS16

Sigma-Aldrich

Mayer′s Hematoxylin Solution

Synonym(s):

(6aS,11bR)-7,11b-Dihydroindeno[2,1-c]chromene-3,4,6a,9,10(6H)-pentol, Delafield′s hematoxylin, Progressive stains

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

1 g/L

technique(s)

microbe id | staining: suitable

pH

2.4 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.

Application

General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.

Other Notes

1 g/L certified hematoxylin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Høybye Bosteen et al.
Atherosclerosis, 265, 93-101 (2017-09-04)
Chronic kidney disease is characterized by uremia and causes premature death, partly due to accelerated atherosclerosis. Apolipoprotein (apo) M is a plasma carrier protein for the lipid sphingosine-1-phosphate (S1P). The Apom-S1P complex associates with HDL, and may contribute to its
Shyambabu Chaurasiya et al.
Cancer gene therapy, 27(3-4), 125-135 (2019-06-19)
Oncolytic viruses have shown excellent safety profiles in preclinical and clinical studies; however, in most cases therapeutic benefits have been modest. We have previously reported the generation of a chimeric poxvirus (CF33), with significantly improved oncolytic characteristics, through chimerization among
Max H Kushner et al.
EMBO reports, 21(1), e48741-e48741 (2019-12-04)
Transcription factors critical for the transition of normal breast epithelium to ductal carcinoma in situ (DCIS) and invasive breast cancer are not clearly defined. Here, we report that the expression of a subset of YAP-activated and YAP-repressed genes in normal mammary
Hanna Kalamarz-Kubiak et al.
Biology open, 6(10), 1493-1501 (2017-09-02)
In this
Franziska Roth-Walter et al.
Scientific reports, 7, 45067-45067 (2017-03-24)
Acrolein, a highly reactive unsaturated aldehyde, is generated in large amounts during smoking and is best known for its genotoxic capacity. Here, we aimed to assess whether acrolein at concentrations relevant for smokers may also exert immunomodulatory effects that could

Questions

1–9 of 9 Questions  
  1. What would be the best solution to dilute product MHS16 to a 50% concentration?

    1 answer
    1. Aluminum-based hematoxylin solutions, including Mayers Hematoxylin, should never be diluted before use. Mayers Hematoxylin solution is ready to use as it comes from the bottle. The issue with diluting aluminum-based hematoxylins is that while the hematoxylin strength is diluted, so is the aluminum concentration. Mayers hematoxylin is a progressive hematoxylin, formulated with enough aluminum to prevent the hematoxylin from nonspecifically binding with the cytoplasm. Diluting the aluminum may lead to staining of the cytoplasm by the hematoxylin. As Mayers Hematoxylin has one of the lower hematoxylin concentrations among popular hematoxylin formulations, the best solution is to simply reduce the amount of time the tissues are stained in the Mayers Hematoxylin.

      Helpful?

  2. Is there an alternative product available for the MHS series of Mayers Hematoxylin solutions?

    1 answer
    1. The Mayers Hematoxylin available is the MHS series of hematoxylin solutions, comeS in various sizes from 100ml to 4L. These solutions are all formulated with chloral hydrate and no alcohol. There is also a Mayers Hemalum solution available as product number 109249, which is formulated with ethylene glycol in place of chloral hydrate. Additionally, there is an Acid Hematoxylin formulated with chloral hydrate, with catalog number 2852-100ml.

      Helpful?

  3. What is the recommended bluing agent for MHS16? Also, what is the recommended range of incubation time for using MHS16?

    1 answer
    1. The protocol for use simply states to blue in running tap water or a dilute alkaline solution. Warm tap water will be slightly more alkaline than cold tap water. Opt to use dilute alkaline solutions if the pH of tap water tends towards the acidic side, as this could cause issues with the tissues bluing quickly and properly. Among these solutions are saturated lithium carbonate solutions or 15 grams per liter of sodium bicarbonate, which can produce a slightly alkaline solution. Another option is Scott's Tap Water Substitute (catalog number S5134). Reagent or ACS grade lithium carbonate and sodium bicarbonate are suitable for use. Bluing with Scott's Tap Water Substitute produced the most stable stained nuclei. As for the recommended staining time, it is 15 minutes. However, Mayers Hematoxylin is a progressive hematoxylin, so if the nuclear staining is not intense enough, the staining time can be increased. Extending the staining time past 15 minutes should not result in overstaining of the cytoplasm, as Mayers Hematoxylin is formulated to prevent this.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. There is also a product listed on the website as Hematoxylin Solution According to Mayer. What is the difference between product 51275 and the various Mayer's Hematoxylin solutions?

    1 answer
    1. The basic formulations are the same. Both include chloral hydrate, although some of the concentrations could vary slightly. Product 51275 is not intended for In Vitro Diagnostic Use. Product 51275 is not CE Marked. The various Mayer's Hematoyxlin solutions are intended For In Vitro Diagnostic Use and are also CE Marked. Product inserts are available for various languages.

      Helpful?

  6. There are several Mayer's Hematoxylins listed on the website, beginning with the letters MHS.  What are the differences?

    1 answer
    1. Products MHS1, MHS16, MHS32, MHS80 and MHS128 are in fact the same product. In this case a unique product number was assigned to each package size. Other than the package size, there are no differences in formulation or use.

      Helpful?

  7. Is chloral hydrate used in the preparation of Mayer's Hematoxylin?  Does this mean the solution is controlled as a DEA Schedule IV product?

    1 answer
    1. Yes, chloral hydrate is included in the formulation. No, the solution is not controlled as a DEA Schedule IV product. Once the choral hydrate is mixed with the remaining chemicals, it is impossible to obtain the chloral hydrate in a purified form.

      Helpful?

  8. The slides were stained with Mayer's Hematoxylin but the nuclei are not blue. Why?

    1 answer
    1. The nuclei will originally appear as the same color as the solution. Only after the slides have been "blued" will the nuclei appear blue. The pH of the solution is acidic. This acidic pH promotes specific staining. However the slides must be washed in an neutral or alkaline solution for the color to shift to blue. Prolonged warm water washes in tap water are often sufficient for the bluing to occur. To shorten the time spent in washing, various bluing agents are often used to shorten the blueing process. Scott's Tap Water Substitute (Product No. S5134) is one such blueing agent.

      Helpful?

  9. The website states that Mayer's Hematoxylin solution is a progressive type hematoxylin. What is a progressive type hematoxylin?

    1 answer
    1. Hematoxylin solutions are generally classified as either progressive or regressive. Progressive formulations will stain only the nucleus. No staining should occur within the cytoplasm. Regressive hematoxylins have higher dye contents and if used for sufficient periods of time will overstain both the nucleus and the cytoplasm. Acid differentiation is required to remove the excessive stain from the cytoplasm and nucleus. This differentiation take practice to master. Too much or too little stain can be removed during differentiation, resulting in poorly stained slides. For users lacking extensive experience with the Hematoxylin and Eosin staining procedure,  progressive hematoxylin formulations are often the better choice.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service