Skip to Content
Merck
All Photos(1)

Documents

HHS80

Sigma-Aldrich

Harris Hematoxylin Solution, Modified

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

7 g/L

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.

Other Notes

7 g/L certified hematoxylin

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhe Wang et al.
Biochimica et biophysica acta, 1792(6), 555-563 (2009-03-14)
The prion protein (PrP) is essential for the pathogenesis of prion disease. PrP has been detected in the cytosol of neurons and transgenic mice expressing PrP in the cytosol (cyPrP) under a pan-neuronal promoter developed rapid cerebellar granule neuron degeneration.
Christina T Teng et al.
PloS one, 6(11), e26961-e26961 (2011-11-11)
Peroxisome proliferator-activated receptor-γ co-activator-1α (PGC-1α) is a member of the transcriptional coactivator family that plays a central role in the regulation of cellular energy metabolism under various physiological stimuli. During fasting, PGC-1α is induced in the liver and together with
Gloria Valdés et al.
Reproductive biology and endocrinology : RB&E, 6, 13-13 (2008-03-29)
The angiogenic and invasive properties of the cytotrophoblast are crucial to provide an adequate area for feto-maternal exchange. The present study aimed at identifying the localization of interrelated angiogenic, hyperpermeability and vasodilator factors in the feto-maternal interface in pregnant guinea-pigs.
Patricia Fernández-Nogueira et al.
Cancer letters, 424, 70-83 (2018-03-20)
Histamine receptor 1 (HRH1) belongs to the rhodopsin-like G-protein-coupled receptor family. Its activation by histamine triggers cell proliferation, embryonic development, and tumor growth. We recently established that HRH1 is up-regulated in basal and human epidermal growth factor receptor 2 (HER2)-enriched
Hideki Ishii et al.
Bioorganic & medicinal chemistry letters, 22(3), 1469-1474 (2012-01-17)
SAR studies for the exploration a novel class of anti-human immunodeficiency virus type 1 (HIV-1) agents based on the hematoxylin structure (1) are described. The systematic deoxygenations of 1 including asymmetric synthesis were conducted to obtain a compound showing high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service