MBD0024
Microbial DNA standard from Thermobifida fusca
Suitable for PCR, sequencing and NGS, 10 ng/μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105500
NACRES:
NA.51
Recommended Products
Quality Level
form
liquid
concentration
10 ng/μL
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
shipped in
ambient
storage temp.
−20°C
General description
Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses.1 Thermobifida fusca genomic DNA microbial standard can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.T. fusca, is a moderately thermophilic gram-positive soil bacterium, that belongs to actinobacteria, with optimum growth at 55-60°C.2,3 The genome of T. fusca has a high GC content (67.5%) 3 and therefore it may serve as a quality marker for the PCR stage in the NGS workflow. It is a major degrader of plant cell walls and has been used as a model organism for the study of secreted, thermostable cellulases. 3 Thermostable enzymes from T.fusca have been reported to replace thermochemical pre- treatments of cellulosic biomass in the process of decomposition to biofuels.4T. fusca, being a soil bacterium, is not likely to be found in the human microbiome. Therefore, T. fusca genomic DNA standard can be used as an internal standard for the NGS workflow; as a negative control for sample preparation, or as a reference DNA for sequencing.
Read here how to use our standards to ensure data integrity for your microbiome research.
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
The genomic DNA is provided at ≥10 ng/μl concentration in TE buffer pH 8.0.It is recommended to avoid freeze thaw cycles of this product.
Suitability
PCR, sequencing, next generation sequencing
Physical form
Liquid -The genomic DNA is provided at ≥10 ng/μl concentration in TE buffer pH 8.0.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service