Skip to Content
Merck
All Photos(2)

Key Documents

MBD0005

Sigma-Aldrich

Microbial DNA standard from Salmonella enterica

Suitable for PCR, sequencing and NGS, 10 ng/μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.24

Quality Level

form

liquid

concentration

10 ng/μL

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses.1 Salmonella enterica genomic DNA microbial standard can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.
Salmonella enterica serotype Typhimurium (also S. Typhimurium) is a rod-shaped gram-negative bacterium of the family Enterobacteriaceae. It is an important food-borne pathogen that in humans causes intestinal inflammation, diarrhea, and the presence of neutrophils in stool samples.2 Growth of S. enterica in the gut lumen causes depletion of the healthy microbial community.3 Infection with Salmonella can induce inflammation and increase the risk for inflammatory bowel disease (IBD)4

Read here how to use our standards to ensure data integrity for your microbiome research.

Application

Suitable for Quantitative standard for PCR, Sequencing and NGS

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, sequencing and NGS
  • Improve Bioinformatics analyses
  • Increases reproducibility
  • Compare results lab to lab

Physical form

Liquid -The genomic DNA is provided at ≥10 ng/μl concentration in TE buffer pH 8.0

Other Notes

Strain: Salmonella enterica subsp. enterica serovar Typhimurium strain

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fecal leukocytes in diarrheal illness.
J C Harris et al.
Annals of internal medicine, 76(5), 697-703 (1972-05-01)
Kim O Gradel et al.
Gastroenterology, 137(2), 495-501 (2009-04-14)
Various commensal enteric and potentially pathogenic bacteria may be involved in the pathogenesis of inflammatory bowel diseases (IBD). We compared the risk of IBD between a cohort of patients with documented Salmonella or Campylobacter gastroenteritis and an age- and gender-matched
Inna Sekirov et al.
Gut microbes, 1(1), 30-41 (2011-02-18)
Gastrointestinal infections involve an interactive tripartite relationship between the invading pathogen, the host, and the host's resident intestinal microbiota. To characterize the host inflammatory response and microbiota alterations during enteric salmonellosis, C57BL/6 mice were pre-treated with a low dose of
J Paul Brooks et al.
BMC microbiology, 15, 66-66 (2015-04-17)
Characterizing microbial communities via next-generation sequencing is subject to a number of pitfalls involving sample processing. The observed community composition can be a severe distortion of the quantities of bacteria actually present in the microbiome, hampering analysis and threatening the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service