Skip to Content
Merck
All Photos(1)

Key Documents

MAK126

Sigma-Aldrich

Bilirubin Assay Kit

sufficient for 180 colorimetric tests

Synonym(s):

Hematoidin Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 180 colorimetric tests

detection method

colorimetric

relevant disease(s)

hematological disorder; gastrointestinal diseases

storage temp.

2-8°C

General description

Bilirubin, also known as hematoidin, is a degradation product formed as a result of heme catabolism in the liver. Bilirubin circulates in the blood stream as either the unconjugated insoluble form (indirect bilirubin) or the soluble glucuronide-conjugated form (direct bilirubin). Conjugated bilirubin moves from the bile canaliculi of the liver to the gall bladder where it is excreted into the small intestine during digestion. High levels of bilirubin can result in jaundice and may indicate liver disease, blood disorders, or blockage of the bile ducts.

Application

Bilirubin Assay Kit has been used to measure the total bilirubin concentration in samples.

Features and Benefits

Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.

Suitability

Suitable for the detection of of both total and conjugated bilirubin in serum and plasma samples.

Principle

This assay, based on the Jendrassik-Grof method, utilizes the reaction of bilirubin with diazotized sulfanilic acid resulting in a colorimetric product measured at 530 nm, proportionate to the bilirubin present in the sample. This assay kit measures both total and conjugated bilirubin.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly J Jasmer et al.
Pigment cell & melanoma research, 33(6), 850-868 (2020-06-20)
Biosynthesis and degradation of heme, an iron-bound protoporphyrin molecule utilized by a wide variety of metabolic processes, are tightly regulated. Two closely related enzymes, heme oxygenase 1 (HMOX1) and heme oxygenase 2 (HMOX2), degrade free heme to produce carbon monoxide
Zhi-Ming Ding et al.
Frontiers in pharmacology, 9, 410-410 (2018-05-17)
Understanding of the temporal changes of hepatic lesions in the progression and regression of non-alcoholic steatohepatitis (NASH) is vital to elucidation of the pathogenesis of NASH, and critical to the development of a strategy for NASH pharmacotherapy. There are challenges
Progression and regression of hepatic lesions in a mouse model of NASH induced by dietary intervention and its implications in pharmacotherapy.
Ding ZM, et al.
Frontiers in Pharmacology, 9, 410-410 (2018)
Bin Dong et al.
Journal of lipid research, 58(2), 350-363 (2016-12-13)
The farnesoid X receptor (FXR) plays critical roles in plasma cholesterol metabolism, in particular HDL-cholesterol (HDL-C) homeostasis. Obeticholic acid (OCA) is a FXR agonist being developed for treating various chronic liver diseases. Previous studies reported inconsistent effects of OCA on
Morgane M Thibaut et al.
Journal of cachexia, sarcopenia and muscle, 12(1), 70-90 (2020-12-23)
Cancer cachexia is a debilitating metabolic syndrome contributing to cancer death. Organs other than the muscle may contribute to the pathogenesis of cancer cachexia. This work explores new mechanisms underlying hepatic alterations in cancer cachexia. We used transcriptomics to reveal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service