Skip to Content
Merck
All Photos(3)

Key Documents

MAK052

Sigma-Aldrich

ALT Activity Assay

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Alanine Aminotransferase Activity Assay Kit, SGPT Activity Assay Kit, Serum Glutamic-Pyruvic Transaminase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GPT(2875)
mouse ... GPT(76282)
rat ... GPT(81670)

General description

Alanine Aminotransferase (ALT), also known as serum glutamic-pyruvic transaminase (SGPT), is a pyridoxal-phosphate-dependent enzyme that catalyzes the reversible transfer of an amino group from alanine to α-ketoglutarate, generating pyruvate and glutamate. ALT is found primarily in liver and serum, but occurs in other tissues as well. Hepatocellular injury often results in an increase of serum ALT levels and serum ALT levels can be used as a marker for liver injury.

Application

ALT Activity Assay kit has been used to determine the activity of alanine aminotransferase (ALT) in serum samples.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of alanine aminotransferase (ALT) activity in biological samples including tissue, cells and serum.

Principle

The ALT Activity Assay Kit provides a simple and direct procedure for measuring ALT activity in a variety of biological samples.

ALT activity is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/ fluorometric (λex = 535/λem = 587 nm) product, proportional to the pyruvate generated. One unit of ALT is defined as the amount of enzyme that generates 1.0 μmole of pyruvate per minute at 37 °C.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Therapeutic Effects of $\alpha$-Mangostin on Thioacetamide-Induced Cirrhosis in Rats
Nilbunga S.
Journal of the Medical Association of Thailand = Chotmaihet Thangphaet, 98(9), S91-S97 (2015)
Brett Palaschak et al.
Molecular therapy. Methods & clinical development, 5, 142-152 (2017-05-10)
Multiple independent adeno-associated virus (AAV) gene therapy clinical trials for hemophilia B, utilizing different AAV serotypes, have reported a vector dose-dependent loss of circulating factor IX (FIX) protein associated with capsid-specific CD8
L Bočkor et al.
Gene therapy, 24(10), 649-660 (2017-08-15)
Adeno-associated virus (AAV) -mediated gene therapy is a promising strategy to treat liver-based monogenic diseases. However, two major obstacles limit its success: first, vector dilution in actively dividing cells, such as hepatocytes in neonates/children, due to the non-integrating nature of
Resolvin D1 and E1 alleviate the progress of hepatitis toward liver cancer in long-term concanavalin A-induced mice through inhibition of NF-KB activity.
Kuang H, et al.
Oncology Reports, 35(1), 307-317 (2016)
Melissa F Brereton et al.
Nature communications, 7, 13496-13496 (2016-11-25)
Insulin secretion from pancreatic β-cells is impaired in all forms of diabetes. The resultant hyperglycaemia has deleterious effects on many tissues, including β-cells. Here we show that chronic hyperglycaemia impairs glucose metabolism and alters expression of metabolic genes in pancreatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service