Skip to Content
Merck
All Photos(3)

Key Documents

M9281

Sigma-Aldrich

3-Methyladenine

autophagy inhibitor

Synonym(s):

3-MA, 6-Amino-3-methylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5
CAS Number:
Molecular Weight:
149.15
Beilstein:
146087
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99% (HPLC)

form

powder

mp

~300 °C (dec.) (lit.)

solubility

DMF: 9.80-10.20 mg/mL, clear, colorless to light yellow

SMILES string

Cn1cnc(N)c2ncnc12

InChI

1S/C6H7N5/c1-11-3-10-5(7)4-6(11)9-2-8-4/h2-3H,7H2,1H3

InChI key

FSASIHFSFGAIJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyladenine (3-MA) is used to inhibit and study the mechanism of autophagy (lysosomal self-degradation) and apoptosis under various conditions. 3-MA inhibits autophagy by blocking autophagosome formation via the inhibition of type III Phosphatidylinositol 3-kinases (PI-3K). For use as an autophagy inhibitor, 3-MA is typically used at a concentration of 5 mM.
3-Methyladenine has been used as an autophagy inhibitor:
  • to determine the expressions of autophagy-related genes (Beclin 1, light chain 3 (LC3) and p62) using the transfected human bone marrow mesenchymal stem cells (BM-MSCs)
  • with cisplatin to study the inhibition of autophagy influenced cisplatin-induced apoptosis in A2780cp cells
  • to study the interplay between colistin-induced autophagy and apoptosis

Standard for detection of nucleic acid methylation during carcinogenesis.

Biochem/physiol Actions

3-Methyladenine (3-MA) is used to inhibit and study the mechanism of autophagy (lysosomal self-degradation) and apoptosis under various conditions. 3-MA inhibits autophagy by blocking autophagosome formation via the inhibition of type III phosphatidylinositol 3-kinases (PI-3K).

Preparation Note

This product is soluble in DMF (10 mg/mL; may require gentle heating). It is also soluble in water, 95% ethanol, or 1 N NaOH at 30 mg/mL with heating, but upon cooling, the product precipitates from solution. Solutions of 3-MA may be prepared in DMSO at 100 mM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingyuan Chen et al.
Shock (Augusta, Ga.), 52(1), 111-121 (2018-10-05)
Sepsis-induced myopathy is a heavy burden for patients during respiratory failure as well as after discharge, which could be characterized with qualitative changes to nAChR in a rat model of sepsis, regulated by NRG-1. Autophagy is an innate immune defense
Heng Wang et al.
Journal of dairy science, 102(9), 8264-8272 (2019-07-01)
Staphylococcus aureus is an important pathogen causing chronic and subclinical mastitis of cows. Autophagy is an important regulatory mechanism that participates in the elimination of invading pathogenic organisms. Here, we hypothesize that autophagy is involved in the process of Staph.
Ruishuang Ma et al.
Cancer biology & therapy, 20(9), 1206-1212 (2019-05-17)
Autophagy plays a complicated role in tumorigenesis, and the effects of autophagy in drug resistance have not been fully known. The aim of this study was to evaluate autophagy activity in lung cancer cells derived from different origins and explore
Xiao Ye et al.
Peptides, 119, 170120-170120 (2019-07-28)
Insulin resistance (IR) is a fundamental pathogenic factor shared by a myriad of metabolic disorders, including obesity and type 2 diabetes. The mechanism of IR is usually accompanied by mitochondrial dysfunction. Irisin has been proposed to act as a hormone
Shuyan Wang et al.
Cell transplantation, 28(6), 775-782 (2019-01-23)
Astrocytes protection and functional regulation are important strategies to protect against neuronal damage caused by ischemia. Activation of the delta opioid receptor (DOR) could reduce astrocytes damage, although the mechanism remains unclear. The present study aimed to test the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service