Skip to Content
Merck
All Photos(3)

Key Documents

M5571

Sigma-Aldrich

Miltefosine

≥98% (perchloric acid titration)

Synonym(s):

Choline hexadecyl phosphate, HePC, Hexadecyl phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H46NO4P
CAS Number:
Molecular Weight:
407.57
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic (organic)

Quality Level

description

zwitterionic

Assay

≥98% (perchloric acid titration)

form

powder

solubility

H2O: 10 mg/mL, clear, colorless

functional group

phospholipid

lipid type

phospholipids

storage temp.

room temp

SMILES string

[O-]P(OCC[N+](C)(C)C)(OCCCCCCCCCCCCCCCC)=O

InChI

1S/C21H46NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-25-27(23,24)26-21-19-22(2,3)4/h5-21H2,1-4H3

InChI key

PQLXHQMOHUQAKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Easy one-pot synthesis of multifunctionalized indole-pyrrole hybrids as a new class of antileishmanial agents.: This study underlines the necessity for innovative treatments amidst growing resistance to established drugs such as miltefosine which is used as a positive control reference, potentially setting a new direction for antileishmanial drug development (Ciccone et al., 2024).

Biochem/physiol Actions

Inhibitor of protein kinase C and of phosphatidylcholine synthesis. Used for the treatment of visceral and cutaneous leishmaniasis. Active against metronidazole-resistant and -susceptible strains of Trichomonas vaginalis

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas P C Dorlo et al.
Antimicrobial agents and chemotherapy, 52(8), 2855-2860 (2008-06-04)
The pharmacokinetics of miltefosine in leishmaniasis patients are, to a great extent, unknown. We examined and characterized the pharmacokinetics of miltefosine in a group of patients with Old World (Leishmania major) cutaneous leishmaniasis. Miltefosine plasma concentrations were determined in samples
Suresh K Tipparaju et al.
Journal of medicinal chemistry, 51(23), 7344-7347 (2008-11-08)
The first synthesis and biological evaluation of antibiotic 31 (A-33853) and its analogues are reported. Initial screening for inhibition of L. donovani, T. b. rhodesiense, T. cruzi, and P. falciparum cultures followed by determination of IC(50) in L. donovani and
Maria V Papadopoulou et al.
Journal of medicinal chemistry, 54(23), 8214-8223 (2011-10-26)
A series of novel 2-nitro-1H-imidazole- and 3-nitro-1H-1,2,4-triazole-based aromatic and aliphatic amines were screened for antitrypanosomal activity and mammalian cytotoxicity by the Drugs for Neglected Diseases initiative (DNDi). Out of 42 compounds tested, 18 3-nitro-1,2,4-triazoles and one 2-nitroimidazole displayed significant growth
Abdullah Alotaibi et al.
Scientific reports, 9(1), 11364-11364 (2019-08-08)
Extracts of 35 samples of European propolis were tested against wild type and resistant strains of the protozoal pathogens Trypanosoma brucei, Trypanosoma congolense and Leishmania mexicana. The extracts were also tested against Crithidia fasciculata a close relative of Crithidia mellificae
Nathan P Wiederhold et al.
Antimicrobial agents and chemotherapy, 57(2), 745-750 (2012-11-21)
Miltefosine is an alkyl phosphocholine with good oral bioavailability and in vitro activity against Cryptococcus species that has gained interest as an additional agent for cryptococcal infections. Our objective was to further evaluate the in vivo efficacy of miltefosine in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service