Skip to Content
Merck
All Photos(3)

Key Documents

M9153

Sigma-Aldrich

Maltohexaose

≥65% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62O31
CAS Number:
Molecular Weight:
990.86
Beilstein:
79666
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

form

powder

concentration

≥65% (HPLC)

impurities

≤8% water (Karl Fischer)

color

light yellow

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)C(O)O[C@@H]6CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C36H62O31/c37-1-7-13(43)14(44)21(51)32(58-7)64-27-9(3-39)60-34(23(53)16(27)46)66-29-11(5-41)62-36(25(55)18(29)48)67-30-12(6-42)61-35(24(54)19(30)49)65-28-10(4-40)59-33(22(52)17(28)47)63-26-8(2-38)57-31(56)20(50)15(26)45/h7-56H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14+,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31?,32-,33-,34-,35-,36-/m1/s1

InChI key

OCIBBXPLUVYKCH-LIGGPISVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Maltohexaose (G6) was used as the acceptor for quantifying 4-α-glucanotransferase activity in a study that assessed discrimination of porcine glycogen debranching enzyme isozymes. It has also been used in a study to investigate the production of a thermophilic maltooligosyl-trehalose synthase.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donatella Cimini et al.
Journal of industrial microbiology & biotechnology, 35(10), 1079-1083 (2008-07-03)
The thermoacidophilic archaeon Sulfolobus solfataricus MT4 encodes a maltooligosyltrehalose synthase (MTS), that catalyzes an intramolecular transglycosylation process converting the glycosidic linkages at the reducing end of dextrins from alpha-1,4 into alpha-1,1. In this research the gene encoding MTS was cloned
Discrimination of Porcine Glycogen Debranching Enzyme Isozymes by the Ratios of Their 4-α-Glucanotransferase and Amylo-α-1,6-glucosidase Activities
Doi, S., et al.
The Journal of Biological Chemistry, 147, 851-856 (2010)
Rivca L Valk-Weeber et al.
Carbohydrate research, 479, 13-22 (2019-05-18)
Glycoproteins are biologically active proteins of which the attached glycans contribute to their biological functionality. Limited data is available on the functional properties of these N-glycans in isolation, without the protein core. Glycan release, typically performed with the PNGase F
Galina Polekhina et al.
Structure (London, England : 1993), 13(10), 1453-1462 (2005-10-12)
AMP-activated protein kinase (AMPK) coordinates cellular metabolism in response to energy demand as well as to a variety of stimuli. The AMPK beta subunit acts as a scaffold for the alpha catalytic and gamma regulatory subunits and targets the AMPK
Patrick Van Gelder et al.
Journal of bacteriology, 184(11), 2994-2999 (2002-05-11)
The lining of the maltodextrin-specific maltoporin (LamB) channel exhibits a string of aromatic residues, the greasy slide, part of which has been shown previously by crystallography to be involved in substrate binding. To probe the functional role of the greasy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service