Skip to Content
Merck
All Photos(2)

Documents

G1253

Sigma-Aldrich

Xanthan gum from Xanthomonas campestris

Synonym(s):

Gum xanthan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Xanthomonas campestris

form

powder

color

faint yellow to beige

storage temp.

room temp

InChI

1S/C13H10O/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-8H,9H2

InChI key

GJCOSYZMQJWQCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Xanthan gum is an anionic polysaccharide composed of a β-(1→4)-D-glucopyranose glucan backbone with side chains of (1→3)-α-D-mannopyranose-(2→1)-β-D-glucuronic acid-(4→1)-β-D-mannopyranose on alternating residues. Approximately half of the terminal mannose residues are 4,6-pyruvated while most of the inner mannose residues are 6-acetylated. Its properties make it a useful matrix component for drug delivery systems. It forms stable drug suspensions in aqueous media and soft gels with locust bean gum or guar gum. Xanthan gum mimics the texture of lipids and is used as a control in experiments where signaling pathways initiated by consumption of lipid-containing reagents are investigated in mice.

Other Notes

A polysaccharide containing glucose, mannose, potassium glucuronate, acetate and pyruvate. Forms a hydrophilic colloid.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Jain et al.
Phytopathology, 109(10), 1811-1819 (2019-05-16)
'Candidatus Liberibacter' spp. are uncultured insect endosymbionts and phloem-limited bacterial plant pathogens associated with diseases ranging from severe to nearly asymptomatic. 'Ca. L. asiaticus', causal agent of Huanglongbing or citrus "greening," and 'Ca. L. solanacearum', causal agent of potato zebra
Yunmei Luo et al.
International journal of biological macromolecules, 144, 967-977 (2019-11-11)
Controlling and delaying the retrogradation of gelatinized rice starch (GRS) can improve the quality of GRS-based foods during storage. Here, we evaluated the influence of corn protein hydrolysates (CPHs), rice protein hydrolysates (RPHs), wheat protein hydrolysates (WPHs), and soybean protein
Anuj Kumar et al.
Materials science & engineering. C, Materials for biological applications, 104, 109983-109983 (2019-09-11)
We report the development of halloysite nanotubes (HNTs)/carboxylated-cellulose nanocrystals (cCNCs) - reinforced and ionically-crosslinked k-carrageenan (k-CG)/xanthan gum (XG) hydrogels. In this study, cCNCs were extracted from microcrystalline cellulose using ammonium persulfate and exhibit 'spindle-like' nanocrystals with approximate diameter of 15-30 nm
An Thi-Binh Nguyen et al.
Food chemistry, 239, 742-750 (2017-09-07)
Dextran or xanthan were used as model exocellular polysaccharides (EPS) to compare the extraction efficiency of EPS from skim milk acid gels using three different protocols. Extraction yields, residual protein concentrations and the macromolecular properties of extracted EPS were determined.
Anuj Kumar et al.
Biofabrication, 12(2), 025029-025029 (2020-02-08)
To achieve a three-dimensional (3D) microenvironment for complex tissue regeneration is a great challenge when developing biomaterials as artificial extracellular matrix (ECM) with properties similar to that of native tissue. Polysaccharide-based hydrogel shows great potential as ECM in the regeneration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service