Skip to Content
Merck
All Photos(1)

Key Documents

M8437

Sigma-Aldrich

Margatoxin

≥99%, recombinant, expressed in E. coli

Synonym(s):

MgTx

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥99%

form

solid

mol wt

4179

storage temp.

−20°C

Amino Acid Sequence

Thr-Ile-Ile-Asn-Val-Lys-Cys-Thr-Ser-Pro-Lys-Gln-Cys-Leu-Pro-Pro-Cys-Lys-Ala-Gln-Phe-Gly-Gln-Ser-Ala-Gly-Ala-Lys-Cys-Met-Asn-Gly-Lys-Cys-Lys-Cys-Tyr-Pro-His

Biochem/physiol Actions

Voltage-gated (Kv1.3) potassium channel blocker.

Other Notes

Major toxin in Centruroides margaritatus (scorpion)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service