Skip to Content
Merck
All Photos(1)

Key Documents

41698

Sigma-Aldrich

Atto 488 NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Atto 488

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (degree of coupling)

manufacturer/tradename

ATTO-TEC GmbH

λ

in methanol: water (1:1) (with 0.1% perchloric acid)

UV absorption

λ: 501-507 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 488 NHS ester is a hydrophilic fluorescent label with high water solubility. The fluorescence activity is excited efficiently at the 480-515nm range. A suitable excitation source for Atto 488 is the 488 nm line of the Argon-Ion laser.

Application

Atto 488 NHS ester is highly suitable for single-molecule detection applications and high-resolution microscopy such as PALM, dSTORM, and STED. In addition, the dye is used in flow cytometry (FACS) and fluorescence in-situ hybridization (FISH) methods.

Features and Benefits

Characteristic features of the Atto 488 NHS ester are:
  • Strong Absorption.
  • High Fluorescence quantum yield.
  • High Photostability.
  • Excellent water solubility.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karuppiah Chockalingam et al.
Scientific reports, 10(1), 2888-2888 (2020-02-23)
Fabs offer an attractive platform for monoclonal antibody discovery/engineering, but library construction can be cumbersome. We report a simple method - Golden Gate assembly with a bi-directional promoter (GBid) - for constructing phage display Fab libraries. In GBid, the constant
Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Muller, P., et al. et al.
Current Microscopy Contributions to Advances in Science and Technology, 1, 3-12 (2012)
Qian Peter Su et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 15036-15046 (2020-06-17)
Mammalian DNA replication is initiated at numerous replication origins, which are clustered into thousands of replication domains (RDs) across the genome. However, it remains unclear whether the replication origins within each RD are activated stochastically or preferentially near certain chromatin
Christiane Iserman et al.
Molecular cell, 80(6), 1078-1091 (2020-12-09)
We report that the SARS-CoV-2 nucleocapsid protein (N-protein) undergoes liquid-liquid phase separation (LLPS) with viral RNA. N-protein condenses with specific RNA genomic elements under physiological buffer conditions and condensation is enhanced at human body temperatures (33°C and 37°C) and reduced
Kin Man Suen et al.
Nature communications, 11(1), 4242-4242 (2020-08-28)
Membraneless organelles are sites for RNA biology including small non-coding RNA (ncRNA) mediated gene silencing. How small ncRNAs utilise phase separated environments for their function is unclear. We investigated how the PIWI-interacting RNA (piRNA) pathway engages with the membraneless organelle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service