Skip to Content
Merck
All Photos(3)

Key Documents

M4773

Sigma-Aldrich

Metaraminol (+)-bitartrate salt

Synonym(s):

(−)-m-Hydroxyphenylpropanolamine bitartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2 · C4H6O6
CAS Number:
Molecular Weight:
317.29
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.C[C@H](N)[C@H](O)c1cccc(O)c1

InChI

1S/C9H13NO2.C4H6O6/c1-6(10)9(12)7-3-2-4-8(11)5-7;5-1(3(7)8)2(6)4(9)10/h2-6,9,11-12H,10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t6-,9-;1-,2-/m01/s1

InChI key

VENXSELNXQXCNT-IJYXXVHRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Metaraminol (+)-bitartrate salt has been used to study the effects of adrenergic agonists in the mouse model of endometriosis.

Biochem/physiol Actions

α-adrenoceptor agonist.
Metaraminol may serve as a false sympathetic transmitter. It is a mixed α - and β-adrenergic agonist.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Tanaka et al.
Experimental neurology, 152(2), 303-306 (1998-08-26)
To verify whether noradrenergic inputs to the subfornical organ (SFO) are involved in the control system of arterial pressure, we investigated the effects of blood pressure changes on noradrealine (NA) release in the SFO and its surrounding sites using microdialysis
Einly Lim et al.
Artificial organs, 36(5), E110-E124 (2012-04-12)
A heart-pump interaction model has been developed based on animal experimental measurements obtained with a rotary blood pump in situ. Five canine experiments were performed to investigate the interaction between the cardiovascular system and the implantable rotary blood pump over
Studies on the interrelationship between the syntheses of noradrenaline and metaraminol
Johnson GE and Pugsley TA
British Journal of Pharmacology, 39(1), 167-174 (1970)
Impact of admixture time and diluent order on metaraminol concentration.
J N Darvall et al.
Anaesthesia and intensive care, 37(5), 859-860 (2009-09-25)
Adenosine-induced cardiopulmonary arrest in a patient with paroxysmal supraventricular tachycardia--a response.
Nigel S Jenkins et al.
The American journal of emergency medicine, 28(4), 526-526 (2010-05-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service