Skip to Content
Merck
All Photos(2)

Documents

M3800

Sigma-Aldrich

A1F Glycan ammonium salt

from Porcine

Synonym(s):

Mannotriose-(fucosyl-di-[N-acetylglucosamine]), mono-sialyl-bis(galactosyl-N-acetylglucosaminyl), NeuNAc(Gal-GlcNAc)2Man3(GlcNAc)2Fuc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C79H131N5O58
CAS Number:
Molecular Weight:
2078.88
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Porcine

Quality Level

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service