Skip to Content
Merck
All Photos(2)

Key Documents

M1050

Sigma-Aldrich

2α-Mannobiose

≥93% (HPLC)

Synonym(s):

α-D-Man-[1→2]-D-Man, 2-O-α-D-Mannopyranosyl-D-mannopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥93% (HPLC)

form

powder

color

white to light yellow

storage temp.

−20°C

SMILES string

OC[C@H]1OC(O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]2O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-6(16)8(18)10(11(20)21-3)23-12-9(19)7(17)5(15)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9+,10+,11?,12-/m1/s1

InChI key

HIWPGCMGAMJNRG-VXSGSMIHSA-N

General description

2α-Mannobiose is a disaccharide.

Application

2α-Mannobiose has been used in a study to assess affinity chromatography with monolithic capillary columns. It has also been used in a study to investigate the influence of disaccharide structure on prebiotic selectivity in vitro.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Luz Sanz et al.
Journal of agricultural and food chemistry, 53(13), 5192-5199 (2005-06-23)
To obtain structure-function information of a range of carbohydrates, which are available only in very small quantities, an in vitro fermentation method using 7 mg of carbohydrate, 0.7 mL of basal medium, and 1% (w/v) of fecal bacteria was validated
Mohamed Bedair et al.
Journal of chromatography. A, 1044(1-2), 177-186 (2004-09-10)
Monolithic capillary columns with surface-immobilized mannan have been introduced for affinity-based micro-column separations by nano-liquid chromatography (nano-LC) and capillary electrochromatography (CEC). Two kinds of polymethacrylate monoliths were prepared, namely poly(glycidyl methacrylate-co-ethylene dimethacrylate) and poly(glycidyl methacrylate-co-ethylene dimethacrylate-co-[2-(methacryloyloxy)ethyl]trimethyl ammonium chloride) to yield
Hui Zhang et al.
Rapid communications in mass spectrometry : RCM, 22(10), 1579-1586 (2008-04-25)
The discrimination of isomeric disaccharides with different linkage types and different monosaccharide residues--glucose (Glc), galactose (Gal), and mannose (Man) at the non-reducing end--was investigated with tandem mass spectrometry (MS/MS) and linear discriminant analysis (LDA). Conventional matrix-assisted laser desorption/ionization (MALDI)-MS has
Christine Henry et al.
mBio, 10(1) (2019-02-14)
Fungal cell wall mannans are complex carbohydrate polysaccharides with different structures in yeasts and molds. In contrast to yeasts, their biosynthetic pathway has been poorly investigated in filamentous fungi. In Aspergillus fumigatus, the major mannan structure is a galactomannan that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service