Skip to Content
Merck
All Photos(1)

Key Documents

M157

Sigma-Aldrich

MEN-10,376

≥95% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H68N12O10
CAS Number:
Molecular Weight:
1081.22
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)C(Cc3ccc(O)cc3)NC(=O)C(N)CC(O)=O)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NC(Cc6c[nH]c7ccccc67)C(=O)NC(CCCCN)C(N)=O

InChI

1S/C57H68N12O10/c1-31(2)50(69-56(78)48(26-35-30-63-43-16-8-5-13-39(35)43)67-53(75)45(23-32-18-20-36(70)21-19-32)65-52(74)40(59)27-49(71)72)57(79)68-47(25-34-29-62-42-15-7-4-12-38(34)42)55(77)66-46(24-33-28-61-41-14-6-3-11-37(33)41)54(76)64-44(51(60)73)17-9-10-22-58/h3-8,11-16,18-21,28-31,40,44-48,50,61-63,70H,9-10,17,22-27,58-59H2,1-2H3,(H2,60,73)(H,64,76)(H,65,74)(H,66,77)(H,67,75)(H,68,79)(H,69,78)(H,71,72)

InChI key

QQHOFZNACVKNHK-UHFFFAOYSA-N

Gene Information

human ... TACR2(6865)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Tyr-Trp-Val-Trp-Trp-Lys-NH2

Biochem/physiol Actions

Potent and selective NK-2 tachykinin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William G Paterson et al.
Gut, 56(10), 1347-1352 (2007-05-26)
Intraluminal acid evokes reflex contraction of oesophageal longitudinal smooth muscle (LSM) and consequent oesophageal shortening. This reflex may play a role in the pathophysiology of oesophageal pain syndromes and hiatus hernia formation. The aim of the current study was to
L Bartho et al.
British journal of pharmacology, 105(4), 805-810 (1992-04-01)
1. The effect of newly developed, receptor-selective tachykinin antagonists (GR 71,251 for NK1 receptors, MEN 10,376 and L 659,877 for NK2 receptors) on noncholinergic transmission to the circular muscle of the guinea-pig ileum has been investigated. 2. In circular muscle
O Löfgren et al.
Burns : journal of the International Society for Burn Injuries, 25(2), 125-129 (1999-04-20)
Recent studies have proposed that activation of the sensory nervous system after thermal injury induces the release of vasoactive neuropeptides, including tachykinins which contribute to the local inflammatory reaction as well as to the nociceptive transmission at the spinal cord
Koichi Tan-No et al.
Brain research, 952(1), 7-14 (2002-10-05)
Intrathecal (i.t.) administration of big dynorphin (1-10 fmol), a prodynorphin-derived peptide consisting of dynorphin A and dynorphin B, to mice produced a characteristic behavioral response, the biting and/or licking of the hindpaw and the tail along with slight hindlimb scratching
B Bianchi et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 12(1), 6-10 (1999-04-03)
Radioligand binding experiments were performed with crude homogenates from normal human skin in order to investigate substance P receptor density. Binding of [3H]substance P ([3H]SP) reached equilibrium after 20 min and was saturable analysis of saturation curves gave a significantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service